Skip to Content
Merck
All Photos(1)

Key Documents

48836

Supelco

cis-1,2-Dichloroethene solution

certified reference material, 2000 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H2Cl2
CAS Number:
Molecular Weight:
96.94
EC Number:
UNSPSC Code:
12000000

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

2000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

InChI

1S/C2H2Cl2/c3-1-2-4/h1-2H/b2-1-

InChI key

KFUSEUYYWQURPO-UPHRSURJSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Timothy E Mattes et al.
FEMS microbiology reviews, 34(4), 445-475 (2010-02-12)
Extensive use and inadequate disposal of chloroethenes have led to prevalent groundwater contamination worldwide. The occurrence of the lesser chlorinated ethenes [i.e. vinyl chloride (VC) and cis-1,2-dichloroethene (cDCE)] in groundwater is primarily a consequence of incomplete anaerobic reductive dechlorination of
Florian Schevenels et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 19(13), 4335-4343 (2013-01-22)
Highly functionalised benzofurans have been prepared from ortho-hydroxyphenones and 1,1-dichloroethylene. The key intermediate, a chloromethylene furan, smoothly rearranged into the corresponding benzofuran carbaldehyde under acidic conditions. Some mechanistic investigations have been performed and several biologically active benzofurans have been synthesised.
C Andrew Ramsburg et al.
Environmental science & technology, 44(23), 9105-9111 (2010-11-09)
Abiotic and biotic reductive dechlorination with chlorinated ethene dense non-aqueous-phase liquid (DNAPL) source zones can lead to significant fluxes of complete and incomplete transformation products. Accurate assessment of in situ rates of transformation and the potential for product sequestration requires
Darlene D Wagner et al.
BMC genomics, 13, 200-200 (2012-05-24)
Geobacter lovleyi is a unique member of the Geobacteraceae because strains of this species share the ability to couple tetrachloroethene (PCE) reductive dechlorination to cis-1,2-dichloroethene (cis-DCE) with energy conservation and growth (i.e., organohalide respiration). Strain SZ also reduces U(VI) to
Federico Aulenta et al.
Bioresource technology, 101(24), 9728-9733 (2010-08-17)
Quinone moieties in humic substances have previously been shown to serve as extracellular electron acceptors in different metabolic pathways. Here we show that the humic acid analogue antraquinone-2,6-disulfonate (AQDS) can also serve as an electron donor in the microbial reductive

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service