Skip to Content
Merck
All Photos(1)

Key Documents

500976

Supelco

Vinyl chloride solution

certified reference material, 2000 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12000000

grade

certified reference material

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

2000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care
petroleum

format

single component solution

storage temp.

2-8°C

SMILES string

ClC=C

InChI

1S/C2H3Cl/c1-2-3/h2H,1H2

InChI key

BZHJMEDXRYGGRV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here


Vinyl chloride, is a regulated and carcinogenic compound, belonging to the class of organochlorides. It is an industrially important chemical, used as a monomer in the production of polyvinyl chloride (PVC), which finds a variety of applications in food wrapping films, raincoats, electric insulation, pipes and bottles, plastic containers, etc.
Vinyl chloride is a colorless gas, which can serve as a monomer for the industrial production of poly(vinyl chloride) homopolymer and copolymer. It can also be used in the production of vinyl stearate, vinylidene chloride, tri- and tetra-chloroethylene. It can be obtained via reaction of hydrochloric acid with acetylene derived from calcium carbide.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Vinyl chloride solution has been used as a certified reference material for the quantification of the analyte in air samples using thermal desorption gas chromatography mass spectrometry (TD-GC−MS).

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 1A - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup - Solvent

Flash Point(C)

11.0 °C - closed cup - Solvent

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vinyl chloride: still a cause for concern.
Kielhorn J, et al.
Environmental Health Perspectives, 108(7), 579-588 (2019)
Intratec Solutions
Vinyl Chloride Production from Ethylene Dichloride - Cost Analysis - VCM E11A (2016)
Intrusion of chlorinated hydrocarbons and their degradation products from contaminated soil. Measurement of indoor air quality and biomonitoring by analysis of end-exhaled air
Scheepers JTP, et al.
The Science of the Total Environment, 653, 223-230 (2019)
J Phillip Kaiser et al.
International journal of toxicology, 31(6), 551-563 (2012-12-01)
Liver disease is a major health issue characterized by several pathological changes, with steatosis (fatty liver) representing a common initial step in its pathogenesis. Steatosis is of critical importance because prevention of fatty liver can obviate downstream pathologies of liver
G M Bodienkova et al.
Meditsina truda i promyshlennaia ekologiia, (3)(3), 21-27 (2013-06-22)
This paper considers the features of disorders of immunity in chronic exposure to vinyl chloride at work and in experimental animals. It is shown that one of the earliest and most sensitive indicators of the formation of the resistance of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service