Skip to Content
Merck
All Photos(1)

Key Documents

254991

Sigma-Aldrich

Trichlorofluoromethane

≥99.5%

Synonym(s):

CFC-11, Fluorotrichloromethane, Freon® 11

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl3CF
CAS Number:
Molecular Weight:
137.37
Beilstein:
1732469
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

5.04 (vs air)

Quality Level

vapor pressure

12.85 psi ( 20 °C)
39.17 psi ( 55 °C)

Assay

≥99.5%

form

liquid

refractive index

n20/D 1.3821 (lit.)

bp

23.7 °C (lit.)

mp

−111-−110 °C (lit.)

solubility

water: soluble 1 g/L

density

1.494 g/mL at 25 °C (lit.)

functional group

chloro
fluoro

storage temp.

2-8°C

SMILES string

FC(Cl)(Cl)Cl

InChI

1S/CCl3F/c2-1(3,4)5

InChI key

CYRMSUTZVYGINF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Rotational absorption spectrum for the ground state of trichlorofluoromethane (Freon-11) has been measured on the part per thousand levels by volume by time-resolved THz spectroscopy. Reductive dehalogenation of trichlorofluoromethane catalyzed by Methanobacterium barkeri has been reported.

Application

Please view www.aldrich.com/epaods regarding the EPA′s request for application information of Ozone Depleting Substances

Legal Information

Freon is a registered trademark of E. I. du Pont de Nemours and Company

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Ozone 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dehalogenation of trichlorofluoromethane (CFC-11) by Methanosarcina barkeri.
Krone UE and Thauer RK.
FEMS Microbiology Letters, 90(2), 201-204 (1992)
Terahertz (THz) spectroscopy of Freon-11 (CCl3F, CFC-11) at room temperature.
Altan H, et al.
Chemical Physics Letters, 427(4), 241-245 (2006)
E Jüngling et al.
Analytical biochemistry, 171(1), 150-157 (1988-05-15)
A one-vial procedure has been developed to quantify free fatty acids in human blood serum and rat heart tissue. To allow routine analysis the system has been constructed to allow simultaneous processing of nine samples (Praechromat). The free fatty acids
A Groppi et al.
Journal of forensic sciences, 39(3), 871-876 (1994-05-01)
A case of lethal poisoning due to trichlorofluoromethane (FC11) inhalation is described. The fluorocarbon was determined in biological tissues by headspace gas chromatography-mass spectrometry. FC11 was detected in all the examined tissues, with decreasing levels in heart, lung, brain, liver
P Sharma et al.
Rapid communications in mass spectrometry : RCM, 18(20), 2383-2387 (2004-09-24)
Nonresonant laser-induced multiphoton dissociation/ionization studies have been conducted for trichloromethane (CHCl3) and trichlorofluoromethane (CFCl3) at 355 nm, using time-of-flight mass spectrometry (TOFMS). The molecular ion signal was found to be missing for both these compounds, and very similar fragmentation patterns

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service