23242-U
Septa, PTFE/Silicone
tan PTFE/silicone septum, septum diam. × total thickness × PTFE thickness 18 mm × 0.060 in. × 2 mil, pkg of 144 ea
Synonym(s):
PTFE-Silicone Composite Septa
About This Item
Recommended Products
material
PTFE face
silicone body
tan PTFE/silicone septum
packaging
pkg of 144 ea
parameter
250 °C temp. limit
septum diam. × total thickness × PTFE thickness
18 mm × 0.060 in. × 2 mil
compatibility
for use with 30 mL (32 × 80 mm) ~ For Bottle (Volume and Dimensions): 60 mL (38 × 95 mm)
for use with products with 20/400 thread
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service