Skip to Content
Merck

Skip To

1379183

USP

Melamine

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

2,4,6-Triamino-1,3,5-triazine, sym-Triaminotriazine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
C3N3(NH2)3
CAS Number:
Molecular Weight:
126.12
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
124341
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

melamine

manufacturer/tradename

USP

mp

>300 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Nc1nc(N)nc(N)n1

InChI

1S/C3H6N6/c4-1-7-2(5)9-3(6)8-1/h(H6,4,5,6,7,8,9)

InChI key

JDSHMPZPIAZGSV-UHFFFAOYSA-N

General description

Find answers to frequently asked questions on USP products in USP FAQs.
This compound is listed in the SVHC (Substances of very high concern) candidate list of ECHA (European Chemicals Agency)
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Melamine USP Reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monograph such as metformin Hydrochloride

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
52549M2659BP1295
description

United States Pharmacopeia (USP) Reference Standard

description

analytical standard

description

99%

description

British Pharmacopoeia (BP) Reference Standard

manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

BP

grade

pharmaceutical primary standard

grade

analytical standard

grade

-

grade

pharmaceutical primary standard

application(s)

pharmaceutical (small molecule)

application(s)

cleaning products
cosmetics
food and beverages
personal care

application(s)

-

application(s)

pharmaceutical

format

neat

format

neat

format

-

format

-

API family

melamine

API family

-

API family

-

API family

-

agency

USP

agency

-

agency

-

agency

BP


Still not finding the right product?

Explore all of our products under Melamine


pictograms

Health hazard

signalword

Warning

Hazard Classifications

Carc. 2 - Repr. 2 - STOT RE 2

target_organs

Urinary tract

Storage Class

11 - Combustible Solids

flash_point_f

572.0 °F - closed cup

flash_point_c

300 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters