Skip to Content
Merck
All Photos(12)

Key Documents

ZRB1557

Sigma-Aldrich

Anti-phospho-GSK3β Ser9 Antibody, clone 6K18 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Glycogen synthase kinase-3 beta

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

6K18, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 46.74 kDa
observed mol wt ~45 kDa

purified by

using Protein A

species reactivity

human, mouse

species reactivity (predicted by homology)

rat

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable
peptide synthesis: suitable
western blot: suitable

dilution

(Peptide Inhibition assay)

isotype

IgG

epitope sequence

N-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

phosphorylation (pSer9)

Gene Information

human ... GSK3B(2932)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 6K18 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects GSK-3b phosphorylated on serine 9.

Immunogen

KLH-conjugated linear peptide corresponding to 9 amino acids surrounding phosphoserine 9 in human GSK-3b.

Application

Quality Control Testing

Evaluated by Western Blotting in lysate from HeLa cells treated with phorbol 12-myristate 13-acetate (PMA).

Western Blotting Analysis (WB): A 1:10,000 dilution of this antibody detected GSK-3β phosphorylated on serine 9 in lysate from HeLa cells treated with phorbol 12-myristate 13-acetate (PMA; 500 nM; 30 min).

Tested applications

Western Blotting Analysis: A 1:10,000 dilution from a representative lot detected phospho-GSK3β Ser9 in lysate from NIH3T3 cells treated with PDGF.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected phospho-GSK3β-Ser9 in HeLa cells treated with phorbol 12-myristate 13-acetate (PMA)..

Peptide Inhibition Assay: Target band detection in lysate from HeLa cell treated with phorbol 12-myristate 13-acetate m(PMA) was prevented by preblocking of a representative lot with the immunogen phosphopeptide, but not the corresponding non-phosphopeptide.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Glycogen synthase kinase-3 beta (UniProt: P49841; also known as EC:2.7.11.26, GSK-3 beta, Serine/threonine-protein kinase GSK3B) is encoded by the GSK3B gene (Gene ID: 2932) in human. GSK-3 is a multifunctional serine/threonine kinase, is a key regulator of numerous signaling pathways during embryogenesis and in metabolic control. Two isoforms of GSK-3 are reported in mammals: a 51 kDa GSK-3a and a 47 kDa GSK-3b. These two isoforms exhibit about 98% homology in their kinase domains but share only about 36% identity in the last 76 C-terminal amino acid residues. GSK-3a contains a glycine-rich extension at its N-terminal. GSK-3 is constitutively active in cells and is regulated through inhibition of its activity. It shows a preference for target proteins that are pre-phosphorylated at a priming residue located C-terminal to the site of GSK-3 phosphorylation. Priming phosphorylation, although not absolutely required, enhances the efficiency of phosphorylation of most GSK-3 substrates. Phosphorylation of a threonine residue in the activation loop (T-loop) is considered to be essential for its activity. The T-loop of GSK-3a is phosphorylated at Tyr279 and GSK-3b at Tyr216, which play a role in forcing open the substrate-binding site of the enzyme. A primary mechanism of GSK3 activity regulation is phosphorylation of N-terminal serine residues (Serine 21 in GSK-3a and serine 9 in GSK-3b). Phosphorylation of these sites is inhibitory to GSK3 kinase activity because the phosphorylated N-terminus acts as a competitive inhibitor for primed substrates. GSK-3b is constitutively active in resting cells and treatment of cells with an agent, such as insulin, causes GSK-3 inactivation through a PI 3-kinase (PI 3-K)-dependent mechanism. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service