Skip to Content
Merck
All Photos(3)

Key Documents

SAB4300001

Sigma-Aldrich

Anti-phospho-GSK3B (pSer9) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-glycogen synthase kinase 3 beta antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~46 kDa

species reactivity

mouse, rat, human

concentration

1 mg/mL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:100
indirect immunofluorescence: 1:100-1:200
western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(T-T-SP-F-A)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer9)

Gene Information

human ... GSK3B(2932)

Immunogen

Peptide sequence around phosphorylation site of serine 9 (T-T-S(p)-F-A), according to the protein GSK3B.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Participates in the Wnt signaling pathway. Implicated in the hormonal control of several regulatory proteins including glycogen synthase, MYB and the transcription factor JUN. Phosphorylates JUN at sites proximal to its DNA-binding domain, thereby reducing its affinity for DNA. Phosphorylates MUC1 in breast cancer cells, and decreases the interaction of MUC1 with CTNNB1/beta-catenin. Phosphorylates CTNNB1/beta-catenin.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yea-Hwey Wang et al.
Food & function, 10(8), 4725-4738 (2019-07-16)
Antrodia camphorata is a well-known traditional Chinese mushroom used as a functional food and nutraceutical in Taiwan and China. The aim of this study was to explore the protective effects and mechanism(s) of the ethyl acetate crude extract of A.
Shayesteh Mehdinejadiani et al.
Biology of reproduction, 100(3), 641-648 (2018-09-06)
Polycystic ovary syndrome (PCOS) is an endocrine disorder in women of reproductive age. In addition to anovulation, endometrial dysfunction can reduce fertility in PCOS. The cyclical changes of endometrium are controlled by estrogen and progesterone via modulating the Wnt/B-catenin pathway.
Xiujing Feng et al.
Journal of cellular physiology, 234(10), 18994-19009 (2019-03-29)
Acute kidney injury (AKI) is a frequent and serious complication of sepsis; however, there are currently no effective therapies. Inflammation and oxidative stress are the major mechanisms implicated in lipopolysaccharide (LPS)-induced AKI. Dexmedetomidine (DEX) has been reported to have remarkable
Pilar Sánchez-Blázquez et al.
The international journal of neuropsychopharmacology, 21(10), 938-948 (2018-06-04)
Several currently available animal models reproduce select behavioral facets of human mania as well as the abnormal glutamatergic neurotransmission and dysregulation of glycogen synthase kinase 3β that accompanies this disease. In this study, we addressed the therapeutic potential of ligands
Jianping Guo et al.
Nature cell biology, 21(2), 226-237 (2019-01-30)
Aberrant activation of AKT disturbs the proliferation, survival and metabolic homeostasis of various human cancers. Thus, it is critical to understand the upstream signalling pathways governing AKT activation. Here, we report that AKT undergoes SETDB1-mediated lysine methylation to promote its

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service