Skip to Content
Merck
All Photos(14)

Key Documents

ZRB1174

Sigma-Aldrich

Anti-Peripherin Antibody, clone 1D4, ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43
conjugate:
unconjugated
application:
ICC
IHC (p)
WB
clone:
1D4, recombinant monoclonal
species reactivity:
mouse, rat, human
citations:
technique(s):
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable (peptide)

biological source

rabbit (recombinant)

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1D4, recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 53.65 kDa
observed mol wt ~53 kDa

species reactivity

mouse, rat, human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable (peptide)

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... PRPH(5630)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 1D4 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects Peripherin. It targets an epitope within 17 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 17 amino acids from the C-terminal region of human Peripherin.

Application

Anti-Peripherin, clone 1D4 ZooMAb , Cat. No. ZRB1174, is a highly specific rabbit recombinant monoclonal antibody that targets Peripherin and has been tested for use in Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected Peripherin in mouse spinal cord tissue lysate.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected Peripherin in human esophagus and rat spinal cord tissue sections.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected Peripherin in PC12 and SH-SY5Y cells.

Note: Actual optimal working dilutions must be determined by the end user as specimens and experimental conditions may vary.

Target description

Peripherin (UniProt: P41219; also known as Neurofilament 4) is encoded by the PRPH (also known as NEF4, PRPH1) gene (Gene ID: 5630) in human. Peripherin is a member of the intermediate filament family. It is classified as a class-III neuronal intermediate filament protein that is expressed mainly in neurons of the peripheral nervous system. It is also found in neurons of the central nervous system that have projections toward peripheral structures, such as spinal motor neurons. Its expression is also reported in certain neuroendocrine tumors. Peripherin is shown to coexist with the neurofilament triplet proteins (neurofilament light, medium, and heavy chain). PRPH gene expression is reported to be the highest in late embryonic and early postnatal brain and declines somewhat before expression of triplet protein sharply rises. A possible peripherin involvement in amyotrophic lateral sclerosis (ALS) pathogenesis has been suggested based on studies with transgenic mice that display over-expression and with a toxic splicing variant of the gene. Two isoforms of peripherin have been described that are produced by alternative splicing. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Leung, CL., et al., (2004). Brain Pathol. 14(3); 290-296; Yuan, A., et al. (2012). J. Neurosci. 32(25); 8501-8508).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose. Normal appearance is a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

30 μg/mL after reconstitution at 25 μL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service