Skip to Content
Merck
All Photos(3)

Key Documents

MAB5262

Sigma-Aldrich

Anti-Neurofilament H (200 kDa) Antibody

CHEMICON®, mouse monoclonal, RT97

Synonym(s):

Anti-CMT2CC, Anti-NFH

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

Product Name

Anti-Neurofilament 200 kDa Antibody, clone RT97, clone RT97, Chemicon®, from mouse

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

RT97, monoclonal

species reactivity

mouse, human

species reactivity (predicted by homology)

rat

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... NEFH(4744)
mouse ... Nefh(380684)
rat ... Nefh(24587)

General description

Neurofilaments are a type of intermediate filament that serve as major elements of the cytoskeleton supporting the axon cytoplasm. They are the most abundant fibrillar components of the axon, being on average 3-10 times more frequent than axonal microtubules. Neurofilaments (10nm in dia.) are built from three intertwined protofibrils which are themselves composed of two tetrameric protofilament complexes of monomeric proteins. The neurofilament triplet proteins (68/70, 160, and 200 kDa) occur in both the central and peripheral nervous system and are usually neuron specific. The 68/70 kDa NF-L protein can self-assemble into a filamentous structure, however the 160 kDa NF-M and 200 kDa NF-H proteins require the presence of the 68/70 kDa NF-L protein to co-assemble. Neuromas, ganglioneuromas, gangliogliomas, ganglioneuroblastomas and neuroblastomas stain positively for neurofilaments. Although typically restricted to neurons, neurofilaments have been detected in paragangliomas and adrenal and extra-adrenal pheochromocytomas. Carcinoids, neuroendocrine carcinomas of the skin, and oat cell carcinomas of the lung also express neurofilaments. For more neurofilament information see Nervous System Cell Type Specific Marker chart online under the Millipore Technical Support section.

Specificity

The antibody recognizes an epitope present on human neurofilaments 200 kDa and both bovine 160 kDa and 200 kDa neurofilaments (2). lt reacts with numerous neurites in human senile plaques and decorates neurofibrillary tangles (NFT) (i.e. on the pyramidal cells of the hippocampus and on neurons of the cerebral cortex of subjects with senile dementia of the Alzheimer’s type).

Immunogen

Rat brain protein preparation

Application

Anti-Neurofilament 200 kDa Antibody, clone RT97 is an antibody against Neurofilament 200 kDa for use in IH, IH(P) & WB.
Immunohistochemistry:
1-10 μg/mL

Immunohistochemistry(paraffin):
Representative images from a previous lot.
Optimal Staining With Citrate Buffer, pH 6.0, Epitope Retrieval: Human Cerebellum

Optimal working dilutions must be determined by end user.
Research Category
Neuroscience
Research Sub Category
Neurofilament & Neuron Metabolism

Neuronal & Glial Markers

Quality

Evaluated by Western Blot on Mouse brain membrane lysates.

Western Blotting Analysis:
1:500 dilution of this antibody detected NEUROFILAMENT on 10 µg of Mouse brain membrane lysates.

Target description

200 kDa

Physical form

Format: Purified
Protein A purified
Purified mouse monoclonal IgG1 in 0.02 M Phosphate buffer, 0.25 M NaCl with 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8ºC from date of receipt.

Analysis Note

Control
Brain

Cultured neurons

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yinhai Wang et al.
PloS one, 7(2), e30632-e30632 (2012-03-01)
Co-localisation is a widely used measurement in immunohistochemical analysis to determine if fluorescently labelled biological entities, such as cells, proteins or molecules share a same location. However the measurement of co-localisation is challenging due to the complex nature of such
S Hou et al.
Neuroscience, 159(1), 369-379 (2009-01-17)
Autonomic dysreflexia is a potentially life-threatening hypertensive syndrome following high thoracic (T) spinal cord injury (SCI). It is commonly triggered by noxious pelvic stimuli below the injury site that correlates with increased sprouting of primary afferent C-fibers into the lumbosacral
H U De Schepper et al.
The Journal of physiology, 586(21), 5247-5258 (2008-08-30)
Patients with inflammatory bowel disease often suffer from gastrointestinal motility and sensitivity disorders. The aim of the current study was to investigate the role of transient receptor potential of the vanilloid type 1 (TRPV1) receptors in the pathophysiology of colitis-induced
Shuhei Ito et al.
eNeuro, 5(5) (2018-12-19)
Nogo receptor-1 (NgR1) signaling is involved in the limitation of axonal regeneration following spinal cord injury (SCI) through collapsing the growth cone and inhibiting neurite outgrowth. Lateral olfactory tract usher substance (LOTUS), a NgR antagonist, suppresses these pathological conditions. A
Meiying Li et al.
Experimental and therapeutic medicine, 15(4), 3929-3939 (2018-03-28)
Bone marrow mesenchymal stem cell (BMSC) transplantation is effective for repairing spinal cord injuries (SCIs); however, there are limitations of clinical BMSC applications. Previously, we reported that dermal papilla cells (DPCs) secrete brain-derived neurotrophic factor and glial cell line-derived neurotrophic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service