Skip to Content
Merck
All Photos(1)

Key Documents

Z665614

Hettich® centrifuge rotors

for ROTOFIX 32A and UNIVERSAL 320/320R Centrifuges, Swing-out Rotor, 8 places

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103900
NACRES:
NB.22

feature

to hold 8 x 50 mL tubes with screw caps

packaging

pack of 1 ea

manufacturer/tradename

Hettich 1617

parameter

2,469 max. RCF

technique(s)

centrifugation: suitable

compatibility

for use with centrifuge models Universal 320/320R and Rotafix 32A

Looking for similar products? Visit Product Comparison Guide

Related Categories

Legal Information

Hettich is a registered trademark of Andreas Hettich GmbH & Co. KG

required but not provided

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service