Z652555
Greiner CELLSTAR® serological pipette
capacity 50 mL , 0.5 mL accuracy
Synonym(s):
pipette, serological pipette, Greiner serological pipettes
About This Item
Recommended Products
material
polystyrene
sterility
sterile; γ-irradiated
sterile
feature
RNase and DNase free
graduations Subdivisions 0.5mL
plugged
packaging
pack of 100 ea
manufacturer/tradename
Greiner 768180
capacity
50 mL , 0.5 mL accuracy
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Precision is aided by fine-line graduation printing providing accuracy of better than ±2%.
Features and Benefits
- 1 mL, 2 mL and 5 mL sizes incorporate drawn tips while larger sizes feature molded tips to provide maximum accuracy
- Packed individually, then double packed for extra protection
- Designed to fit all standard pipetting devices
- Cotton plugged for safety
- Sterilized by γ-irradiation
- DNase, RNase & human DNA free
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service