Key Documents
Z623504
Hettich® plate carriers
for ROTINA 38/38R and UNIVERSAL 320/320R, plate carrier for rotors 1645; 1725
About This Item
UNSPSC Code:
41103910
NACRES:
NB.22
feature
Holds 1 x MTP, CPplate
packaging
pack of 4 ea
manufacturer/tradename
Hettich 4345
technique(s)
centrifugation: suitable
compatibility
for use with rotors 1645 (Z727938) & 1725 (Z631124)
General description
  • compatible with rotors 1645 and 1725 (Z631124)
Legal Information
Hettich is a registered trademark of Andreas Hettich GmbH & Co. KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)