Key Documents
Z327085
BRAND® BLAUBRAND® volumetric flask, glass stopper, amber glass
volume 250 mL, accuracy: 0.15 mL
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
material
borosilicate glass 3.3 flask
Joint
joint ST/NS neck14/23
feature
class A DE-M, DIN EN ISO 1042. Calibrated to contain (TC, In).
packaging
pack of 2 ea
manufacturer/tradename
BRAND 37451
volume measuring range
250 mL, accuracy: 0.15 mL
capacity
250 mL
General description
Volumetric flasks according to DIN EN ISO 1042, Class A, conformity certified. Made from Boro 3.3, calibrated to contain (TC, IN). Supplied with a interchangeable amber glass stopper. Marks and inscriptions in high contrast white enamel.
All BLAUBRAND® volumetric flasks are supplied with a batch number and an accompanying batch certificate in the original packaging.
Other Notes
BR37451 (India)
Legal Information
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG