Skip to Content
Merck
All Photos(2)

Documents

WH0010461M1

Sigma-Aldrich

Monoclonal Anti-MERTK antibody produced in mouse

clone 2D2, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-MER, Anti-c-mer proto-oncogene tyrosine kinase, Anti-cmer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2D2, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MERTK(10461)

General description

The gene MERTK (MER proto-oncogene, tyrosine kinase) is mapped to human chromosome 2q14.1. The encoded protein belongs to the Tyro3, Axl, and Mertk (TAM) family of receptor tyrosine kinases. MERTK is strongly expressed in macrophages, ovary, prostate, testis, lung and kidney. The protein has a conserved intracellular kinase domain and an extracellular adhesion molecule-like domain.

Immunogen

MERTK (NP_006334, 21 a.a. ~ 120 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
AITEAREEAKPYPLFPGPFPGSLQTDHTPLLSLPHASGYQPALMFSPTQPGRPHTGNVAIPQVTSVESKPLPPLAFKHTVGHIILSEHKGVKFNCSINVP

Biochem/physiol Actions

MERTK (MER proto-oncogene, tyrosine kinase) is mainly involved in the starting of efferocytosis, which is the removal of dying cells by phagocytosis. It interacts with two ligands, Gas6 (growth arrest-specific 6) and Protein-S. Mutation in this gene regulates the severity of fibrosis in patients with non-alcoholic fatty liver disease. Mutations in MERTK gene also result in autosomal recessive retinal degeneration due to faults in phagocytosis. MERTK is upregulated in malignant cells, including leukemia, lymphoma and colorectal cancer. Variants of this gene are detected in melanoma, multiple myeloma, renal cancer and carcinoma. MERTK is also overexpressed in sclerotic lesions.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A novel start codon mutation of the MERTK gene in a patient with retinitis pigmentosa.
Jinda W
Molecular Vision, 342-351 (2016)
Human iPSC derived disease model of MERTK-associated retinitis pigmentosa.
Lukovic D, et.al
Scientific Reports, 5, 12910-12910 (2015)
Andreea Cristina Mihaila et al.
Cells, 13(3) (2024-02-09)
Following myocardial infarction (MI), blood neutrophils quickly and extensively infiltrate the heart, where they are temporally polarized into pro-inflammatory (N1) and anti-inflammatory (N2) subpopulations. Neutrophil transmigration is rapidly followed by the accrual of macrophages (MACs), which are believed to undergo
MERTK rs4374383 polymorphism affects the severity of fibrosis in non-alcoholic fatty liver disease.
Petta S
Journal of Hepatology, 64(3), 682-690 (2016)
A Comprehensive Review of Mutations in the MERTK Proto-Oncogene.
Parinot C and Nandrot EF
Advances in Experimental Medicine and Biology, 854, 259-265 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service