Skip to Content
Merck
All Photos(1)

Key Documents

SAB4301137

Sigma-Aldrich

Anti-ACTIN antibody produced in rabbit

affinity isolated antibody

Synonym(s):

ACTB

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

42 kDa

species reactivity

plant

concentration

1 mg/mL

technique(s)

western blot: 1:500-1:2000 (Cell Lysate)

isotype

IgG

accession no.

NP_190236.1

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Arabidopsis thaliana ... ACT12(823805)

General description

Actin/ actin β(ACTB) is encoded by the gene mapped to human chromosome 7p15-p12. In mammals, actin protein consists of six different isoforms encoded by separate genes. The four isoforms αskeletal-actin, αcardiac-actin, αsmooth-actin, and γ smooth-actin are expressed in skeletal, cardiac and smooth muscle. The other two isoforms, β cyto-actin and γ cyto-actin are ubiquitously expressed.

Specificity

The antibody detects endogenous levels of total ACTIN protein.

Immunogen

Synthetic peptide corresponding to residues near the N terminal of Arabidopsis thaliana actin

Biochem/physiol Actions

Actin plays a vital role in various cellular processes, such as cell division, migration, junction formation, chromatin remodeling, transcriptional regulation, vesicle trafficking, and cell shape regulation. Polymorphism in the smooth muscle α-actin (ACTA2) is associated with the development of thoracic aortic aneurysms and dissections.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in pH7.3 PBS, 0.05% NaN3, 50% Glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elizabeth Feldeverd et al.
Frontiers in plant science, 11, 610052-610052 (2021-01-16)
Plants adapt to heat via thermotolerance pathways in which the activation of protein folding chaperones is essential. In eukaryotes, protein disulfide isomerases (PDIs) facilitate the folding of nascent and misfolded proteins in the secretory pathway by catalyzing the formation and
Sublocalization of an invasion-inducing locus and other genes on human chromosome 7.
Habets G G M, et al.
Cytogenetic and genome research, 60(3-4), 200-205 (1992)
Dong Meng et al.
The Plant journal : for cell and molecular biology, 106(5), 1278-1297 (2021-03-19)
Calcineurin B-like (CBL)-interacting protein kinases (CIPKs) play a central role in Ca2+ signalling and promote drought tolerance in plants. The CIPK gene family in pigeon pea (Cajanus cajan L.), a major food crop affected by drought, has not previously been
Lucie Horakova et al.
Frontiers in physiology, 13, 1033130-1033130 (2023-01-27)
Ectodysplasin (Eda) plays important roles in both shaping the developing tooth and establishing the number of teeth within the tooth row. Sonic hedgehog (Shh) has been shown to act downstream of Eda and is involved in the initiation of tooth
Kimihito Saito et al.
Tissue barriers, 11(3), 2106113-2106113 (2022-07-28)
Lipolysis-stimulated lipoprotein receptor (LSR), a lipid metabolism-related factor localized in tricellular tight junctions (tTJs), plays an important role in maintaining the epithelial barrier. LSR is highly expressed in well-differentiated endometrial endometrioid carcinoma (EEC), and its expression decreases during malignancy. Angubindin-1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service