Skip to Content
Merck

Skip To

V6629

Vitamin B12

BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%

Synonym(s):

α-(5,6-Dimethylbenzimidazolyl)cyanocobamide, CN-Cbl, Cyanocob(III)alamin, Cyanocobalamin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C63H88CoN14O14P
CAS Number:
Molecular Weight:
1355.37
PubChem Substance ID:
eCl@ss:
34058004
UNSPSC Code:
12352205
NACRES:
NA.75
EC Number:
200-680-0
Beilstein/REAXYS Number:
4122889
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic ((organic))

Quality Segment

product line

BioReagent

assay

≥98%

form

powder

mol wt

Mw 1355.37 g/mol

technique(s)

cell culture | insect: suitable, cell culture | mammalian: suitable, cell culture | plant: suitable

color

red to brown

pH

6

solubility

water: soluble 50 mg/mL, dark red

application(s)

advanced drug delivery

storage temp.

2-8°C

SMILES string

C/C(C1=N[C@]([C@]([C@H](CC(N)=O)[C@@]/2(C)CCC(NC[C@@H](OP([O-])(O[C@H]3[C@@H](O)[C@@H](N4C=[N]5C6=C4C=C(C)C(C)=C6)O[C@@H]3CO)=O)C)=O)([H])N([Co+]5C#N)C2=C(C)/C([C@@H](CCC(N)=O)C/7(C)C)=NC7=C/8)(C)[C@@](C)(CC(N)=O)[C@@H]1CCC(N)=O)=C9N=C8[C@@H](CCC(N)=O)[C@

InChI

1S/C62H90N13O14P.CN.Co/c1-29-20-39-40(21-30(29)2)75(28-70-39)57-52(84)53(41(27-76)87-57)89-90(85,86)88-31(3)26-69-49(83)18-19-59(8)37(22-46(66)80)56-62(11)61(10,25-48(68)82)36(14-17-45(65)79)51(74-62)33(5)55-60(9,24-47(67)81)34(12-15-43(63)77)38(71-55)23-42-58(6,7)35(13-16-44(64)78)50(72-42)32(4)54(59)73-56;1-2;/h20-21,23,28,31,34-37,41,52-53,56-57,76,84H,12-19,22,24-27H2,1-11H3,(H15,63,64,65,66,67,68,69,71,72,73,74,77,78,79,80,81,82,83,85,86);;/q;;+2/p-2/t31-,34-,35-,36-,37+,41-,52-,53-,56-,57?,59-,60+,61+,62+;;/m1../s1

InChI key

RMRCNWBMXRMIRW-QJRSUKKJSA-L

General description

Cyanocobalamin (B12) is a water soluble vitamin, commonly called as cobalamins. Dairy products, meat and eggs are the major sources of this vitamin.

Application

Vitamin B12 has been used:
  • as a component of Erdschreiber′s medium
  • as a supplement of Dulbecco′s modified Eagle′s medium used for the isolation of ventricular myocytes from neonatal and adult rats
  • orally administered to male quails to study its role in reducing the negative effects of sodium selenite intoxication, in vivo
  • as a supplement in DMEM/ medium 199 (M199) medium for differentiation of human myoblasts.

Biochem/physiol Actions

Vitamin B12 is metabolized to its active form, methylcobalamin. Hindered vitamin B12 uptake is observed in pernicious anemia. Vitamin B12 is essential for erythroblast development. Pregnancy reduces the levels of vitamin B12. Aging reduces vitamin B12 absorption. It works as a coenzyme in fat, carbohydrate and protein metabolism. It is required for growth, genetic stability and survival of cells in vitro. Functions to support one-carbon metabolism. Present in many classical and serum-free formulations.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
6791V2876BP466
description

BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%

description

-

description

≥98%

description

British Pharmacopoeia (BP) Assay Standard

technique(s)

cell culture | plant: suitable, cell culture | insect: suitable, cell culture | mammalian: suitable

technique(s)

-

technique(s)

HPLC: suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

assay

≥98%

assay

≥95% dry basis (UV)

assay

≥98%

assay

-

biological source

synthetic ((organic))

biological source

-

biological source

-

biological source

-

application(s)

advanced drug delivery

application(s)

-

application(s)

-

application(s)

pharmaceutical (small molecule)

form

powder

form

powder

form

powder

form

solid

storage temp.

2-8°C

storage temp.

15-25°C

storage temp.

2-8°C

storage temp.

2-8°C


Still not finding the right product?

Explore all of our products under Vitamin B12


Storage Class

11 - Combustible Solids

wgk

WGK 1

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters