Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Vitamin B
12
Synonym(s):
α-(5,6-Dimethylbenzimidazolyl)cyanocobamide, CN-Cbl, Cyanocob(III)alamin, Cyanocobalamin
Empirical Formula (Hill Notation):
C
63
H
88
CoN
14
O
14
P
CAS Number:
68-19-9
Molecular Weight:
1355.37
EC Number:
200-680-0
Products
Product Category
vitamins (2)
Brand
Sigma-Aldrich (2)
Biological Source
Color
Formula Weight
Physical Form
Product Line
Purity
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
V2876
Vitamin B
12
,
≥98%
Expand
View Pricing
V6629
Vitamin B
12
,
BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Expand
View Pricing