Skip to Content
Merck
All Photos(1)

Key Documents

V108

Sigma-Aldrich

(±)-Norverapamil hydrochloride

≥98% (HPLC)

Synonym(s):

N-Nor-(±)-verapamil hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H36N2O4 · HCl
CAS Number:
Molecular Weight:
477.04
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 15 mg/mL, clear

SMILES string

CC(C)C(CCCNCCC1=CC=C(OC)C(OC)=C1)(C#N)C2=CC(OC)=C(OC)C=C2.Cl

InChI

1S/C26H36N2O4.ClH/c1-19(2)26(18-27,21-9-11-23(30-4)25(17-21)32-6)13-7-14-28-15-12-20-8-10-22(29-3)24(16-20)31-5;/h8-11,16-17,19,28H,7,12-15H2,1-6H3;1H

InChI key

OEAFTRIDBHSJDC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Norverapamil is an N-demethylated metabolite of verapamil. It has significantly less calcium channel blocking ability and exhibit therapeutic activity.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J M Dethy et al.
Journal of chromatography. B, Biomedical applications, 654(1), 121-127 (1994-03-18)
An analytical method has been developed to determine simultaneously the verapamil and norverapamil enantiomers in human plasma using capillary electrophoresis. Among the cyclodextrins tested as chiral selector, only trimethyl-beta-cyclodextrin was suitable to resolve the four enantiomers. The analysis was achieved
Joong-Hwa Chung et al.
Biopharmaceutics & drug disposition, 30(2), 90-93 (2009-02-20)
Verapamil is known to be a P-glycoprotein (P-gp) substrate and norverapamil is formed via hepatic cytochrome P450 (CYP 3A) in the rat. Epigallocatechin gallate (EGCG), a flavonoid, was reported to be an inhibitor of both P-gp and CYP3A. Hence, it
Shan-Shan Xie et al.
The Journal of pharmacy and pharmacology, 62(4), 440-447 (2010-07-08)
The aim was to investigate the effect of Huang-Lian-Jie-Du-Decoction (HLJDD) on the pharmacokinetic behaviour of verapamil in rats. Rats orally received 3.33 g/kg of HLJDD extract for 14 days, and pharmacokinetics of verapamil was investigated after oral and intravenous verapamil.
Shuijun Li et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 847(2), 174-181 (2006-11-23)
A simple, accurate and selective LC-MS/MS method was developed and validated for simultaneous quantification of ten antiarrhythic drugs (diltiazem, amiodarone, mexiletine, propranolol, sotalol, verapamil, bisoprolol, metoprolol, atenolol, carvedilol) and a metabolite (norverapamil) in human plasma. Plasma samples were simply pretreated
Marc-Etienne Castella et al.
Pharmaceutical research, 23(7), 1543-1553 (2006-06-17)
The aims of the study are to develop and evaluate an in vitro rat intestine segmental perfusion model for the prediction of the oral fraction absorbed of compounds and to assess the ability of the model to study intestinal metabolism.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service