Skip to Content
Merck
All Photos(1)

Documents

T8951

Sigma-Aldrich

Tangeretin

≥95% (HPLC)

Synonym(s):

4′,5,6,7,8-Pentamethoxyflavone, 5,6,7,8-Tetramethoxy-2-(4-methoxyphenyl)-4H-1-benzopyran-4-one, NSC 53909, NSC 618905, Ponkanetin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H20O7
CAS Number:
Molecular Weight:
372.37
Beilstein:
351695
EC Number:
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Assay

≥95% (HPLC)

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

SMILES string

COc1ccc(cc1)C2=CC(=O)c3c(OC)c(OC)c(OC)c(OC)c3O2

InChI

1S/C20H20O7/c1-22-12-8-6-11(7-9-12)14-10-13(21)15-16(23-2)18(24-3)20(26-5)19(25-4)17(15)27-14/h6-10H,1-5H3

InChI key

ULSUXBXHSYSGDT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tangeretin is a polymethoxyflavone extracted primarily from the peels of citrus fruits like Citrus sinensis and Citrus reticulata.

Application

Tangeretin has been used as an antioxidant and antifungal agent to test its effects on ferroptosis inhibition, conidial development, and appressorium formation against Magnaporthe oryzae. It has also been used as a co-chemotherapeutics agent to study its cytotoxic effects in combination with 5-Fluorouracil (5-Fu) on WiDr colon cancer cells.

Biochem/physiol Actions

Tangeretin displays its anti-cancer effects by modulating different pathways in several cancer types in both in-vitro and in-vivo studies. It also exerts antioxidant, anti-inflammatory, and estrogenic properties.
Tangeretin is a flavonoid found in the peel of citrus fruits where it most likely provides natural resistance to fungi. Tangeretin has been shown to counteract tumor promoter-induced inhibition of intercellular communication and to inhibit cell proliferation in several cancer lines.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Combination of Tangeretin and 5-Fluorouracil Modulates Cell Cycle and Induce Apoptosis on WiDr Cells
Indriyani L, et al.,
Indonesian Journal of Cancer Chemoprevention, 3(1), 364-369 (2012)
Jérôme Quintin et al.
Bioorganic & medicinal chemistry letters, 19(1), 167-169 (2008-11-18)
A series of chalcones polyoxygenated on the ring A (with pentamethoxy or 2'-hydroxy-3',4',5',6'-tetramethoxy substitution patterns) was synthesized from tangeretin, a natural Citrus flavonoid. These chalcones were evaluated for their antiproliferative, activation of apoptosis, inhibition of tubulin assembly and antileishmanial activities.
Hideo Satsu et al.
Journal of agricultural and food chemistry, 56(13), 5366-5373 (2008-06-11)
The pregnane X receptor (PXR) is understood to be the key regulator for gene expression of such drug-metabolizing enzymes and transporters as multidrug-resistant protein 1 (MDR1) and the cytochrome P450 (CYP) family. We examined the effect of dietary phytochemicals on
Myung Sunny Kim et al.
Molecular and cellular endocrinology, 358(1), 127-134 (2012-04-06)
Although the flavonoid tangeretin (5, 6, 7, 8, 4'-pentamethoxyflavone) is known to possess beneficial health effects, the anti-diabetic effects and the mechanism of action have not been elucidated. Treatment with 100 μM tangeretin significantly increased the uptake of 2-NBDG in
Juhee Seo et al.
European journal of pharmacology, 673(1-3), 56-64 (2011-11-02)
Tangeretin, a natural polymethoxylated flavone concentrated in the peel of citrus fruits, is known to have antiproliferative, antiinvasive, antimetastatic and antioxidant activities. However, the effect of tangeretin on vascular smooth muscle cells (VSMCs) is unknown. This study examined the effect

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service