Skip to Content
Merck

Skip To

1467848

USP

Nobiletin

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

2-(3,4-Dimethoxyphenyl)-5,6,7,8-tetramethoxy-4H-1-benzopyran-4-one, 3′,4′,5,6,7,8-Hexamethoxyflavone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C21H22O8
CAS Number:
Molecular Weight:
402.39
NACRES:
NA.24
UNSPSC Code:
41116107
Beilstein/REAXYS Number:
360887
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

packaging

pkg of 100 mg

manufacturer/tradename

USP

application(s)

pharmaceutical

format

neat

storage temp.

−20°C

SMILES string

COc1ccc(cc1OC)C2=CC(=O)c3c(OC)c(OC)c(OC)c(OC)c3O2

InChI

1S/C21H22O8/c1-23-13-8-7-11(9-15(13)24-2)14-10-12(22)16-17(25-3)19(26-4)21(28-6)20(27-5)18(16)29-14/h7-10H,1-6H3

InChI key

MRIAQLRQZPPODS-UHFFFAOYSA-N

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Nobiletin USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Bitter Orange Young Fruit
  • Bitter Orange Young Fruit Powder
  • Tangerine Peel
  • Tangerine Peel Dry Extract
  • Tangerine Peel Powder

Biochem/physiol Actions

Nobiletin suppresses the expression of matrix metalloproteinases 1, 3 and 9, which participate in the break down of the extracellular matrix during tumor metastasis. Nobiletin is also involved in the increased expression of tissue inhibitor of matrix metalloproteinase 1 further preventing extracellular matrix degradation during tumor invasion. This bioflavonoid is believed to achieve both of these functions by disrupting the signal transduction pathway of Ras-Raf-MEK.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHL80858PHL82477N1538
manufacturer/tradename

USP

manufacturer/tradename

PhytoLab

manufacturer/tradename

PhytoLab

manufacturer/tradename

-

grade

pharmaceutical primary standard

grade

primary reference standard

grade

primary reference standard

grade

-

format

neat

format

neat

format

neat

format

-

application(s)

pharmaceutical

application(s)

food and beverages

application(s)

food and beverages

application(s)

-

packaging

pkg of 100 mg

packaging

-

packaging

-

packaging

-

agency

USP

agency

-

agency

-

agency

-


Still not finding the right product?

Explore all of our products under Nobiletin


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters