Skip to Content
Merck
All Photos(4)

Key Documents

T7576

Sigma-Aldrich

Anti-TGN46 antibody produced in rabbit

enhanced validation

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-MGC14722, Anti-TGN38 homolog, Anti-TGN48, Anti-TGN51, Anti-TGOLN2, Anti-TTGN2, Anti-Trans-Golgi network integral membrane protein 2 precursor, Anti-Trans-Golgi network protein TGN51, Anti-Trans-golgi network protein 2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 80-100 kDa

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

concentration

~1 mg/mL

technique(s)

immunoprecipitation (IP): 10-20 μg using TGN46 from human HepG2 cell lysates.
indirect immunofluorescence: 5-10 μg/mL using human A549 cells
western blot (chemiluminescent): 0.25-0.5 μg/mL using extracts of human HEK-293T cells expressing recombinant human TGN46

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TGOLN2(10618)

General description

Anti-TGN46 recognizes human trans-golgi network protein 2 (TGN46). Detection of the TGN46 band by immunoblotting is specifically inhibited by the immunizing peptide.
TGN46 (Trans-Golgi network protein, 46 kDa), the human homologue of rat TGN38, is a resident integral membrane protein of the trans-Golgi network (TGN) that cycles constitutively between the TGN and the plasma membrane, returning via endosomes. The TGN is the major sorting compartment of the secretory pathway for proteins, lipids and membrane traffic. TGN46 is a heavily glycosylated protein, probably involved in regulating membrane traffic to and from the TGN. TGN46 contains a signal peptide, lumenal domain, membrane-spanning domain, and cytoplasmic domain. The membrane spanning region and cytoplasmic tail contain the retention and retrieval signals, respectively for localization in the TGN. TGN46 is widely expressed.

Immunogen

synthetic peptide corresponding to amino acid residues 426-437 of human TGN46 with N-terminal added cysteine, conjugated to KLH. The corresponding sequence is identical in monkey and differs by three amino acids in rat and mouse.

Application

Rabbit polyclonal anti-TGN46 antibody is used to tag trans-golgi network protein 2 for detection and quantitation by immunocytochemical and immunohistochemical (IHC) techniques such as immunoblotting, immunoprecipitation, and immunofluorescence. It is used as a probe to determine the presence and roles of trans-golgi network protein 2 as a membrane traffic regulating component of the trans-Golgi network (TGN). Anti-TGN46 may be used as a TGN marker.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiehua Li et al.
Autophagy, 13(2), 345-358 (2016-11-23)
The membrane source for autophagosome biogenesis is an unsolved mystery in the study of autophagy. ATG16L1 forms a complex with ATG12-ATG5 (the ATG16L1 complex). The ATG16L1 complex is recruited to autophagic membranes to convert MAP1LC3B-I to MAP1LC3B-II. The ATG16L1 complex
Yong Yu et al.
The Biochemical journal, 434(3), 383-390 (2011-01-22)
Peptide hormones and neuropeptides are packaged and stored in a specialized intracellular organelle called the dense core vesicle. It remains elusive how peptide cargoes are correctly sorted. In the present study, we show that a highly conserved Golgi-localized protein named
Andrea N Dietz et al.
Journal of virology, 92(1) (2017-10-20)
The human cytomegalovirus (HCMV) tegument protein pUL71 is required for efficient secondary envelopment and accumulates at the Golgi compartment-derived viral assembly complex (vAC) during infection. Analysis of various C-terminally truncated pUL71 proteins fused to enhanced green fluorescent protein (eGFP) identified
Christine M Calton et al.
PLoS pathogens, 13(5), e1006200-e1006200 (2017-05-04)
The human papillomavirus type 16 (HPV16) L2 protein acts as a chaperone to ensure that the viral genome (vDNA) traffics from endosomes to the trans-Golgi network (TGN) and eventually the nucleus, where HPV replication occurs. En route to the nucleus
Na Dong et al.
Nature microbiology, 2, 16236-16236 (2016-12-13)
Legionella pneumophila, the causative bacterium for Legionnaires' disease, hijacks host membrane trafficking for the maturation of the Legionella-containing vacuole (LCV). The LCV membrane mainly contains PtdIns4P, which is important for anchoring many secreted Legionella effectors onto the LCV. Here, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service