Skip to Content
Merck
All Photos(1)

Key Documents

SAB4200585

Sigma-Aldrich

Anti-ERGIC-53 antibody, Mouse monoclonal

clone ERGIC-3, purified from hybridoma cell culture

Synonym(s):

Monoclonal Anti-ER-Golgi intermediate compartment 53 kDa protein, Monoclonal Anti-ERGIC-53 antibody produced in mouse, Monoclonal Anti-ERGIC53, Monoclonal Anti-F5F8D, Monoclonal Anti-Gp58, Monoclonal Anti-Intracellular mannose-specific lectin MR60, Monoclonal Anti-LMAN1, Monoclonal Anti-Lectin mannose-binding 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

ERGIC-3, monoclonal

form

buffered aqueous solution

mol wt

antigen ~58 kDa

species reactivity

mouse, human, rat

concentration

~1 mg/mL

technique(s)

immunoblotting: 1-2 μg/mL using whole extracts of human HeLa cells.

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LMAN1(3998)
mouse ... Lman1(70361)
rat ... Lman1(116666)

General description

Endoplasmic reticulum-Golgi intermediate compartment 53kDa protein (ERGIC-53), a nonglycosylated, hexameric type I integral membrane protein is also known as LMAN1 (lectin, mannose-binding 1). This gene is located on human chromosome 18q21.32.

Immunogen

synthetic peptide corresponding to an internal sequence of human ERGIC-53, conjugated to KLH. The corresponding sequence is identical in mouse, rat, bovine and monkey.

Biochem/physiol Actions

Endoplasmic reticulum-Golgi intermediate compartment protein 53kDa protein (ERGIC-53) plays an important role in the proliferation of arenavirus, coronavirus and filovirus. This intracellular cargo receptor enables the anterograde passage of very few glycoprotein ligands in the early exocytic pathway. Mutations in ERGIC-53 (LMAN1) gene results in the deficiency of factor V and factor VIII.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Two new mutations at ERGIC-53 gene in a Turkish family
Torun, et al.
Clinical and Applied Thrombosis/Hemostasis : Official Journal of the International Academy of Clinical and Applied Thrombosis/Hemostasis, 17(3), 248-250 (2011)
Morihisa Fujita et al.
The Journal of cell biology, 194(1), 61-75 (2011-07-06)
Glycosylphosphatidylinositol (GPI) anchoring of proteins is a posttranslational modification occurring in the endoplasmic reticulum (ER). After GPI attachment, proteins are transported by coat protein complex II (COPII)-coated vesicles from the ER. Because GPI-anchored proteins (GPI-APs) are localized in the lumen
Dependence on electron transport chain function and intracellular signaling of genomic responses in SH-SY5Y cells to the mitochondrial neurotoxin MPP+
Brill L B II, et al.
Experimental Neurology, 181(1), 25-38 (2003)
The intracellular cargo receptor ERGIC-53 is required for the production of infectious arenavirus, coronavirus, and filovirus particles
Klaus JP, et al.
Cell host & microbe, 14(5), 522-534 (2013)
Xiang Gui et al.
Nature, 567(7747), 262-266 (2019-03-08)
Cyclic GMP-AMP (cGAMP) synthase (cGAS) detects infections or tissue damage by binding to microbial or self DNA in the cytoplasm1. Upon binding DNA, cGAS produces cGAMP that binds to and activates the adaptor protein STING, which then activates the kinases IKK

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service