Skip to Content
Merck
All Photos(2)

Key Documents

T6031

Sigma-Aldrich

DL-Thiorphan

≥98% (TLC)

Synonym(s):

(±)-N-(3-Mercapto-2-benzylpropionyl)glycine, DL-3-Mercapto-2-benzylpropanoylglycine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H15NO3S
CAS Number:
Molecular Weight:
253.32
Beilstein:
4872101
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (TLC)

form

powder

mp

138 °C

solubility

ethanol: soluble 49.00-51.00 mg/mL, clear, colorless

SMILES string

OC(=O)CNC(=O)C(CS)Cc1ccccc1

InChI

1S/C12H15NO3S/c14-11(15)7-13-12(16)10(8-17)6-9-4-2-1-3-5-9/h1-5,10,17H,6-8H2,(H,13,16)(H,14,15)

InChI key

LJJKNPQAGWVLDQ-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Thiorphan, also called DL-mercapto-2-benzylpropanoyl-glycine is a specific and potent inhibitor of enkephalinase in nanomolar concentrations. It is an inhibitor of neutral endopeptidase. It interacts with cluster of differentiation 10 (CD10) in bone marrow cells and may modulate hematopoiesis. Thiorphan is a safe drugprescribed for diarrhea. It prevents cortical damage in newborn mice and is a potent agent for treating cerebral palsy.(CP)

Application

DL-Thiorphan has been used to evaluate the activity of NEP-dependent neutral endopeptidase. DL-Thiorphan has also been used for determining NEP enzymatic activity using fluorimetric assays.

Biochem/physiol Actions

Enkephalinase inhibitor.

Features and Benefits

This compound is featured on the Neuropeptidases page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Preparation Note

DL-Thiorphan is soluble in ethanol at 49.00 - 51.00 mg/ml and yields a clear, colorless solution.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Global brain delivery of neprilysin gene by intravascular administration of AAV vector in mice
Iwate N, et al.
Scientific Reports (2013)
Thiorphan, an inhibitor of neutral endopeptidase/enkephalinase (CD10/CALLA) enhances cell proliferation in bone marrow cultures of patients with acute leukemia in remission
Stanovic S, et al.
Haematologia, 30(1), 1-10 (2000)
Alexander W Sauter et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 60(3), 393-399 (2018-07-14)
Patients with metastatic medullary thyroid cancer (MTC) have limited systemic treatment options. The use of radiolabeled gastrin analogs targeting the cholecystokinin-2 receptor (CCK2R) is an attractive approach. However, their therapeutic efficacy is presumably decreased by their enzymatic degradation in vivo.
The enkephalinase inhibitor thiorphan shows antinociceptive activity in mice
Roques BP, et al.
Nature, 288(5788), 286-288 (1980)
Methionine enkephalin suppresses metabolic activity of a leukemic cell line (NALM-1) and enhances CD10 expression
Martin-Kleiner I, et al.
International Immunopharmacology, 3, 707-711 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service