Skip to Content
Merck
All Photos(3)

Key Documents

T4425

Sigma-Aldrich

(6R)-5,6,7,8-Tetrahydrobiopterin dihydrochloride

≥98% (HPLC), powder

Synonym(s):

(6R)-BH4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H15N5O3 · 2HCl
CAS Number:
Molecular Weight:
314.17
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

(6R)-5,6,7,8-Tetrahydrobiopterin dihydrochloride,

Quality Level

solubility

water, oxygen free: soluble 19.60-20.40 mg/mL, clear to slightly hazy, colorless to faintly yellow

storage temp.

−20°C

SMILES string

Cl.Cl.C[C@H](O)[C@H](O)[C@H]1CNC2=C(N1)C(=O)NC(N)=N2

InChI

1S/C9H15N5O3.2ClH/c1-3(15)6(16)4-2-11-7-5(12-4)8(17)14-9(10)13-7;;/h3-4,6,12,15-16H,2H2,1H3,(H4,10,11,13,14,17);2*1H/t3-,4+,6-;;/m0../s1

InChI key

RKSUYBCOVNCALL-NTVURLEBSA-N

Gene Information

human ... PAH(5053)

General description

Tetrahydrobiopterin is a natural cofactor for phenylalanine hydroxylase, tyrosine hydroxylase, tryptophan hydroxylase, nitric oxide synthase and alkylglycerol monooxygenase.[1]

Application

(6R)-BH4 has been used as a buffer component for assaying tyrosine hydroxylase enzyme activity in mouse brain tissues. (6R)-BH4 has also been identified as an activator compound from a LOPAC library screen using a fluorogenic α-glucosidase assay.

Biochem/physiol Actions

Lack of tetrahydrobiopterin may cause hyperphenylalaninemia. Diseases like, Parkinson′s, autism, depression and Alzheimer′s shows low concentration of tetrahydrobiopterin in the cerebrospinal fluid.[2] This coenzyme participates in dopamine (DA) synthesis.[3]

Features and Benefits

This compound is featured on the Dopamine, Norepinephrine and Epinephrine Synthesis and Nitric Oxide Synthases pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Preparation Note

(6R)-5,6,7,8-Tetrahydrobiopterin dihydrochloride ((6R)-BH4 ) is soluble in oxygen free water at 19.60 - 20.40 mg/ml and yields a clear to slightly hazy, colorless to faint yellow solution.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kamil Kostyn et al.
Antioxidants (Basel, Switzerland), 9(8) (2020-08-14)
Catecholamines are biogenic aromatic amines common among both animals and plants. In animals, they are synthesized via tyrosine hydroxylation, while both hydroxylation or decarboxylation of tyrosine are possible in plants, depending on the species, though no tyrosine hydroxylase-a counterpart of
Tetrahydrobiopterin biosynthesis as a potential target of the kynurenine pathway metabolite xanthurenic acid
Haruki H, et al.
The Journal of Biological Chemistry (2016)
Ailing Fu et al.
Brain research, 1352, 208-213 (2010-07-06)
Glial-derived neurotrophic factor (GDNF) is a trophic factor for the nigra-striatal tract in experimental Parkinson's disease (PD). The neurotrophin must be administered by intra-cerebral injection, because GDNF does not cross the blood-brain barrier (BBB). In the present study, GDNF was
Keun-Sung Lee et al.
The Korean journal of physiology & pharmacology : official journal of the Korean Physiological Society and the Korean Society of Pharmacology, 17(1), 89-97 (2013-02-27)
Developing an animal model for a specific disease is very important in the understanding of the underlying mechanism of the disease and allows testing of newly developed new drugs before human application. However, which of the plethora of experimental animal
Rebecca Ebenhoch et al.
Proceedings of the National Academy of Sciences of the United States of America, 117(50), 31838-31849 (2020-11-25)
Guanosine triphosphate (GTP) cyclohydrolase I (GCH1) catalyzes the conversion of GTP to dihydroneopterin triphosphate (H2NTP), the initiating step in the biosynthesis of tetrahydrobiopterin (BH4). Besides other roles, BH4 functions as cofactor in neurotransmitter biosynthesis. The BH4 biosynthetic pathway and GCH1

Articles

Serotonin is stored in cells and metabolized by MAO, influencing CNS, GI, and platelet functions.

Serotonin is stored in cells and metabolized by MAO, influencing CNS, GI, and platelet functions.

Serotonin is stored in cells and metabolized by MAO, influencing CNS, GI, and platelet functions.

Serotonin is stored in cells and metabolized by MAO, influencing CNS, GI, and platelet functions.

Questions

1–2 of 2 Questions  
  1. Is the product T4425 (CAS 69056-38-8) soluble in distilled water?

    1 answer
    1. The product is advised to be dissolved in anoxic water. The use of distilled water is not recommended, as it can absorb more oxygen from the atmosphere.

      Helpful?

  2. How should a solution of (6R)-5,6,7,8-tetrahydrobiopterin (hydrochloride) be stored? How long is it good for?

    1 answer
    1. Tetrahydrobiopterin reacts with oxygen, particularly in neutral and alkaline solutions. The product should be reconstituted in oxygen-free water. Tetrahydrobiopterin solutions become yellow but are stable for several weeks at -20°C when prepared at 20-40 mg/mL in 0.1 N HCl.

      Please review the product data sheet for more information on this product:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/277/129/t4425pis.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service