Skip to Content
Merck
All Photos(3)

Key Documents

T2020

Sigma-Aldrich

(±)-α-Tocopherol phosphate disodium salt

water-soluble α-tocopherol analog

Synonym(s):

Alpha Tocopherol phosphate disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C29H49O5PNa2
CAS Number:
Molecular Weight:
554.65
MDL number:
UNSPSC Code:
12352205
eCl@ss:
34058016
PubChem Substance ID:
NACRES:
NA.79

biological source

synthetic (organic)

Quality Level

Assay

≥97% (HPLC)

form

powder

technique(s)

HPLC: suitable

color

white to off-white

storage temp.

2-8°C

SMILES string

[Na].CC(C)CCCC(C)CCCC(C)CCCC1(C)CCc2c(C)c(OP(O)(O)=O)c(C)c(C)c2O1

InChI

1S/C29H51O5P.Na.H/c1-20(2)12-9-13-21(3)14-10-15-22(4)16-11-18-29(8)19-17-26-25(7)27(34-35(30,31)32)23(5)24(6)28(26)33-29;;/h20-22H,9-19H2,1-8H3,(H2,30,31,32);;

InChI key

QRLCTLIPEFQSSG-UHFFFAOYSA-N

Application

(±)-α-Tocopherol phosphate disodium salt has been used as an antioxidant, to assess its protection against oxidative damage in rat hippocampal neurons. It has also been used to study its molecular mechanisms and transport across the plasma membrane.

Biochem/physiol Actions

Tocopherols (TCP) (vitamin E) are a series (α, β, γ and δ) of chiral organic molecules that vary in their degree of methylation of the phenol moiety of the chromanol ring. Tocopherols are lipid soluble anti-oxidants that protect cell membranes from oxidative damage. α-Tocopherol is the form of tocopherol preferentially absorbed by Homo sapiens. α-Tocopherol phosphate (TocP) is used along the α-tocopherol acetate, α-tocopherol-nicotinate, α-tocopherol succinate and other esterified forms as a vitamin E supplement with similar but differential activities.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hitoshi Masaki et al.
Free radical research, 36(6), 705-709 (2002-08-16)
alpha-Tocopherol is a lipophilic vitamin that exhibits an antioxidative activity. The purpose of this study was to clarify the roles of alpha-tocopherol in the regulation of intracellular glutathione (GSH) levels in HaCaT keratinocytes. When HaCaT keratinocytes were cultivated with alpha-tocopherol
Marc Birringer
Molecular nutrition & food research, 54(5), 588-598 (2010-02-20)
Vitamin E is known as the most important lipid antioxidant and is widely used to prevent age-associated diseases. Despite increasing knowledge about human vitamin E metabolism, little is known to justify its widespread use. As meta-analyses revealed even harmful effects
Keiko Nishio et al.
Free radical biology & medicine, 50(12), 1794-1800 (2011-03-30)
α-Tocopheryl phosphate (α-TP), a water-soluble analogue of α-tocopherol, is found in humans, animals, and plants. α-TP is resistant to both acid and alkaline hydrolysis and may exert its own function in this form in vivo. In this study, the uptake
Robert Gianello et al.
Free radical biology & medicine, 39(7), 970-976 (2005-09-06)
We have detected alpha-tocopheryl phosphate in biological tissues including liver and adipose tissue, as well as in a variety of foods, suggesting a ubiquitous presence in animal and plant tissue. Alpha-tocopheryl phosphate is a water-soluble molecule that is resistant to
A transient treatment of hippocampal neurons with alpha-tocopherol induces a long-lasting protection against oxidative damage via a genomic action
de Jesus Ferreira MC, et al.
Free Radical Biology & Medicine (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service