Skip to Content
Merck
All Photos(1)

Key Documents

T1949

Sigma-Aldrich

Total Protein Reagent

Synonym(s):

Biuret Reagent

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NC.07

Quality Level

Application

Total Protein Reagent can be used for the quantitative, colorimetric determination of total protein concentration in solution at 540 nm. The total protein reagent is supplied as a ready-to-use liquid.
Total Protein Reagent has been used in the biuret assay.

Biochem/physiol Actions

A biuret-based total protein reagent. Copper ions in the alkaline reagent react with peptide bonds in proteins resulting in a purple color with an absorbance maximum of 540 nm.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Stefano Cecchini et al.
Animals : an open access journal from MDPI, 10(11) (2020-11-07)
Total antioxidant capacity (TAC) in healthy and dexamethasone-stressed hens was measured by applying four different spectrophotometric methods-the ferric reducing ability (FRAP) assay, the 2,2'-azino-bis (3-ethylbenzotiazoline-6-sulphonic acid) (ABTS) radical cation decolorization assay, the free radical scavenging activity (FRSA), and the total
B T Doumas et al.
Clinical chemistry, 27(10), 1642-1650 (1981-10-01)
We developed a candidate Reference Method for measuring total serum protein by use of the biuret reaction. The method involves a previously described biuret reagent (Clin. Chem. 21: 1159, 1975) and Standard Reference Material (SRM) 927 bovine albumin (National Bureau
Determination of serum proteins by means of the biuret reagent.
Gornall, A. G., et al.,
The Journal of Biological Chemistry, 177, 759-759 (1949)
Chad L Seewagen et al.
Conservation physiology, 8(1), coaa037-coaa037 (2020-05-12)
Non-native, invasive plants can impact birds by altering food sources, nesting substrates and other critical resources. Japanese barberry (Berberis thunbergii) is one of the most invasive, non-native woody plants in in the northeastern USA, and yet almost nothing is known
Does Exurban Housing Development Affect the Physiological Condition of Forest-Breeding Songbirds? A Case Study of Ovenbirds (Seiurus aurocapillus) in the Largest Protected Area in the Contiguous United States.
Seewagen CL
Physiological and Biochemical Zoology : Pbz, 88, 416-424 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service