Skip to Content
MilliporeSigma

Skip To

TP0100

Total Protein Kit, Micro

Synonym(s):

Bradford

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 kit
Check Cart for Availability
$393.00

About This Item

NACRES:
NA.32
UNSPSC Code:
12352202

$393.00


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

wavelength 595 nm
total test time 2 min
working reagent stability 18-26°C for 2 weeks
dynamic range 50 mg/dL 0.5 g/L

Quality Segment

parameter

18-26 °C temperature, 50 μL sample volume

storage temp.

2-8°C

General description

Protein determination is one of the most common operations performed in biochemical research. Total protein kit, micro provides a simple, rapid, and highly sensitive method requiring only 50 μL or less sample.

Application

Colorimetric, Endpoint
Suitable for protein determination for samples of up to 0.5 mg/ml.
Total protein kit, micro has been used to determine:
  • total protein in the bacterial homogenate by Bradford method
  • total protein content by the Bradford microassay from the tissue extracts[1]
  • serum total protein
  • soluble protein content by the Bradford method from leaves

Biochem/physiol Actions

Brilliant blue G (Coomassie blue) reacts with protein in an acid-alcohol medium to form a blue-colored protein dye complex. The color, measured at 595 nm, is proportional to the protein concentration.
Total protein kit, micro can be used to determine the concentration of proteins in solution.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
TP0200PROTTOTPE0230
storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

Quality Level

100

Quality Level

200

Quality Level

200

Quality Level

200

parameter

18-26 °C temperature, 50 μL sample volume

parameter

18-26 °C temperature, 200 μL sample volume

parameter

-

parameter

-

description

wavelength 595 nm
total test time 2 min
working reagent stability 18-26°C for 2 weeks
dynamic range 50 mg/dL 0.5 g/L

description

wavelength 500-750nm

description

-

description

-


Kit Components Also Available Separately

Product No.
Description
SDS & Pricing

  • Protein Standard 5 mL
    SDS

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 3 - Met. Corr. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

flash_point_f

132.8 °F

flash_point_c

56 °C

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters