Skip to Content
Merck
All Photos(1)

Key Documents

T0938

Sigma-Aldrich

Anti-Tumor Necrosis Factor-α antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-TNF-α

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
51111800
NACRES:
NA.41

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

mouse

technique(s)

capture ELISA: suitable
flow cytometry: 5-25 μg/mL
indirect immunofluorescence: 15 μg/mL using mouse cells and tissues
neutralization: suitable
western blot: 0.1-0.2 μg/mL

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Tnf(21926)

Looking for similar products? Visit Product Comparison Guide

General description

Tumor Necrosis Factor-α (TNF-α) is a protein produced by lipopolysaccharide-stimulated macrophages and causes tumor necrosis. It also facilitates many functions like stimulate growth of human fibroblasts and other cell lines, activate polymorphonuclear neutrophils and osteoclasts and collagenase production. Anti-Tumor Necrosis Factor-α antibody can be used for neutralizing the biological activity of recombinant mouse TNF-α. It can also be used in indirect immunofluorescence. Goat anti-Tumor Necrosis Factor-α antibody reacts specifically with recombinant mouse TNF-α.

Specificity

The antibody neutralizes the biological activity of recombinant mouse TNF-α.

Immunogen

recombinant mouse TNF-α

Application

Anti-Tumor Necrosis Factor-α antibody can be used in ELISA capture and flow cytometry. It can also be used in immunoblotting.

Physical form

Lyophilized from 0.2 μm filtered solution in phosphate buffered saline with 5% trehalose.

Preparation Note

Purified using mouse TNF-α affinity chromatography.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E A Carswell et al.
Proceedings of the National Academy of Sciences of the United States of America, 72(9), 3666-3670 (1975-09-01)
In studying "hemorrhagic necrosis" of tumors produced by endotoxin, it was found that the serum of bacillus Calmette--Guerin (BCG)-infected mice treated with endotoxin contains a substance (tumor necrosis factor; TNF) which mimics the tumor necrotic action of endotoxin itself. TNF-positive
J M Dayer et al.
The Journal of experimental medicine, 162(6), 2163-2168 (1985-12-01)
Cachectin/TNF (tumor necrosis factor), an endotoxin-induced murine macrophage hormone implicated in the pathogenesis of cachexia and shock, has been found capable of stimulating collagenase and prostaglandin E2 (PGE2) production by isolated human synovial cells and dermal fibroblasts. This bioactivity associated
D R Bertolini et al.
Nature, 319(6053), 516-518 (1986-02-06)
When leukocytes are exposed to mitogens or antigens in vitro, they release bone-resorbing activity into the culture supernatants which can be detected by bioassay. Like many lymphocyte-monocyte products, this activity has been difficult to purify because of its low abundance
B J Sugarman et al.
Science (New York, N.Y.), 230(4728), 943-945 (1985-11-22)
Modulation of the growth of human and murine cell lines in vitro by recombinant human tumor necrosis factor-alpha (rTNF-alpha) and recombinant human interferon-gamma (rIFN-gamma) was investigated. rTNF-alpha had cytostatic or cytolytic effects on only some tumor cell lines. When administered
Jih-Hua Wei et al.
International journal of molecular sciences, 24(6) (2023-03-30)
Bariatric surgery reduces body weight, enhances metabolic and diabetic control, and improves outcomes on obesity-related comorbidities. However, the mechanisms mediating this protection against cardiovascular diseases remain unclear. We investigated the effect of sleeve gastrectomy (SG) on vascular protection in response

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service