Skip to Content
Merck
All Photos(1)

Key Documents

T0203

Sigma-Aldrich

TGFβR2 (190-end), active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

AAT3, FAA3, HPNCC6, MFS2, RIIC, TAAD2, TGFR-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

Quality Level

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

6-8 nmol/min·mg

mol wt

~68 kDa

UniProt accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... TGFBR2(7048)

General description

Transforming growth factor β receptor 2 (TGFBR2) is a member of the TGFß receptor subfamily and is a serine/threonine protein kinase. The gene encoding this protein is localized on human chromosome 3p24.1.

Biochem/physiol Actions

Transforming growth factor β receptor 2 (TGFBR2)-induced protein phosphorylation plays a key role in signal transduction that leads to mitogenic responses. The TGFBR2 receptor transmits signals from the cell surface to the nucleus and provides instructions for making transforming growth factor (TGF)-β type II receptor. Mutations in TGFBR2 gene have been associated with Marfan syndrome, Loeys-Deitz aortic aneurysm syndrome and the development of various types of tumors.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

TGFBR2-dependent alterations of exosomal cargo and functions in DNA mismatch repair-deficient HCT116 colorectal cancer cells
Fabia Fricke
Cell communication and signaling : CCS (2017)
Loeys?Dietz syndrome: a primer for diagnosis and management
Gretchen
Genitourinary Medicine null
TGF? signalling in context
Joan Massague
Nature Reviews in Molecular and Cell Biology (2012)
Molecular modeling and molecular dynamic simulation of the effects of variants in the TGFBR2 kinase domain as a paradigm for interpretation of variants obtained by next generation sequencing
Michael T
PLoS ONE (2017)
Arthrogryposis as neonatal presentation of Loeys-Dietz syndrome due to a novel TGFBR2 mutation.
Valenzuela I
European Journal of Medical Genetics (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service