SRP3073
IL-12 human
recombinant, expressed in CHO cells, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Synonym(s):
CTL maturation factor (TCMF),, Cytotoxic lymphocyte maturation factor (CLMF), NKSF, TSF
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352202
NACRES:
NA.32
Recommended Products
biological source
human
recombinant
expressed in CHO cells
Assay
≥98% (HPLC)
≥98% (SDS-PAGE)
form
lyophilized
potency
≤1.0 ng/mL ED50
mol wt
40 kDa
packaging
pkg of 10 μg
technique(s)
cell culture | mammalian: suitable
impurities
<0.1 EU/μg endotoxin, tested
color
white to off-white
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... IL12A(3592)
General description
IL-12 (interleukin 12) is produced by activated monocytes/macrophage cells, B lymphocytes and connective tissue type mast cells. Recombinant Human IL-12 is a 75kDa heterodimeric glycoprotein consisting of disulfide-linked 35kDa (p35) and 40kDa (p40) subunits (503 total amino acid residues).
Biochem/physiol Actions
IL-12 (interleukin 12) is a proinflammatory cytokine and plays an important role in protection against pathogens. IL-12 is a potent regulator of cell mediated immune responses and it induces IFN (interferon)-γ production by NK (natural killer) and T cells. Among its biological activities IL-12 promotes the growth and activity of activated NK, CD4+ and CD8+ cells and induces the development of IFN-γ producing Th1 (T helper) cells. Polymorphisms in the IL-12 gene are associated with risk of rheumatoid arthritis. It also exhibits anti-tumor activities by enhancing Th1 adaptive immunity and cytotoxic T lymphocyte functions.
Sequence
p35 Subunit: RNLPVATPDP GMFPCLHHSQ NLLRAVSNML QKARQTLEFY PCTSEEIDHE DITKDKTSTV EACLPLELTK NESCLNSRET SFITNGSCLA SRKTSFMMAL CLSSIYEDLK MYQVEFKTMN AKLLMDPKRQ IFLDQNMLAV IDELMQALNF NSETVPQKSS LEEPDFYKTK IKLCILLHAF RIRAVTIDRV MSYLNAS p40 Subunit: IWELKKDVYV VELDWYPDAP GEMVVLTCDT PEEDGITWTL DQSSEVLGSG KTLTIQVKEF GDAGQYTCHK GGEVLSHSLL LLHKKEDGIW STDILKDQKE PKNKTFLRCE AKNYSGRFTC WWLTTISTDL TFSVKSSRGS SDPQGVTCGA ATLSAERVRG DNKEYEYSVE CQEDSACPAA EESLPIEVMV DAVHKLKYEN YTSSFFIRDI IKPDPPKNLQ LKPLKNSRQV EVSWEYPDTW STPHSYFSLT FCVQVQGKSK REKKDRVFTD KTSATVICRK NASISVRAQD RYYSSSWSEW ASVPCS
Physical form
Lyophilized from 0.5x PBS, pH 7.2.
Reconstitution
Centrifuge the vial prior to opening. Reconstitute in 1x PBS, pH 7.2-7.4 to a concentration of 0.2 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
An absolute and restricted requirement for IL-12 in natural killer cell IFN-gamma production and antiviral defense. Studies of natural killer and T cell responses in contrasting viral infections.
Orange JS and Biron CA
Journal of Immunology, 156, 1138-1142 (1996)
Up-regulation of IL-12 expression in patients with chronic hepatitis B is mediated by the PI3K/Akt pathway.
Wang HW, te al.
Molecular and Cellular Biochemistry, 407, 135-142 (2015)
IL12 polymorphisms, HBV infection and risk of hepatocellular carcinoma in a high-risk Chinese population.
Liu L, et al.
International Journal of Cancer. Journal International Du Cancer, 128, 1692-1696 (2011)
Martin J Page et al.
British journal of haematology, 180(1), 110-117 (2017-11-17)
Inflammation, with its associated inflammatory molecules, is integral to most chronic diseases, including the various cardiovascular diseases. Interleukin 12 (IL12) is one of the inflammatory cytokines that is upregulated during inflammation; however, we know very little about its exact effect
IL-12 induces the production of IFN-gamma by neonatal human CD4 T cells.
Wu CY, et al.
Journal of Immunology, 151, 1938-1949 (1993)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service