SRE0026
Phosphatase substrate
Suitable for manufacturing of diagnostic kits and reagents
Synonym(s):
Phosphatase substrate, 4-Nitrophenyl phosphate disodium salt hexahydrate, pNPP disodium salt hexahydrate
About This Item
Recommended Products
Quality Level
Assay
≥97% (HPLC)
form
powder
solubility
water: 100 mg/mL, clear to slightly hazy
application(s)
diagnostic assay manufacturing
storage temp.
−20°C
SMILES string
O.O.O.O.O.O.[Na+].[Na+].[O-][N+](=O)c1ccc(OP([O-])([O-])=O)cc1
InChI
1S/C6H6NO6P.2Na.6H2O/c8-7(9)5-1-3-6(4-2-5)13-14(10,11)12;;;;;;;;/h1-4H,(H2,10,11,12);;;6*1H2/q;2*+1;;;;;;/p-2
InChI key
KAKKHKRHCKCAGH-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Other Notes
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service