Skip to Content
Merck
All Photos(1)

Key Documents

SNC50

Sigma-Aldrich

mirPremier® microRNA Isolation Kit

1 sufficient for 50 preparations

Synonym(s):

microRNA Isolation Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105501
NACRES:
NA.52

usage

1 sufficient for 50 preparations

technique(s)

DNA extraction: suitable

Looking for similar products? Visit Product Comparison Guide

General description

Sigma′s mirPremier microRNA Isolation Kit provides a rapid and efficient method for purifying and enriching miRNAs and other small RNAs from diverse biological sources, including mammalian cell cultures, animal tissues, plant tissues, and microbial cultures, without using hazardous organic extractions. microRNAs (miRNAs) are a class of small RNA molecules, about 21 nucleotides (nt) in length, that regulate gene expression in a variety of manners, including translational repression, mRNA cleavage and deadenylation. In addition, the kit also can be used for isolating total RNA if messenger RNA or other large RNAs are of interest.
.

Application

mirPremier® microRNA Isolation Kit has been used to:

  • extract total RNA containing miRNA from grape edible plant derived exosome-like nanoparticles (EPDEN).
  • isolate small RNA from S.sirkka and S. napiecek, and S. arctica.
  • extract miRNA, from frozen rat livers for miRNA analysis.

Features and Benefits

  • Designed to enhance the efficiency of isolating microRNA and other small RNA molecules directly from a wide range of biological sources.
  • Enables fast and efficient extraction and concentration of miRNA in 30 minutes for downstream applications.
  • Can extract high-purity miRNA with no detectable large RNA
  • No dangerous organic extractions are involved.

Legal Information

mirPremier is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Interspecies communication between plant and mouse gut host cells through edible plant derived exosome-like nanoparticles.
Mu J
Molecular Nutrition And Food Research, 58(7), 1561-1573 (2014)
Dongxiao Su et al.
Food & function, 8(2), 808-815 (2017-01-26)
Dietary phenolics exhibit hypolipidemic activity by changing lipid metabolism-related microRNA (miRNA) expression. Quercetin 3-O-rutinoside-7-O-α-l-rhamnosidase (quercetin 3-rut-7-rha), rutin and (-)-epicatechin are the main phenolics in lychee (Litchi chinensis Sonn.) pulp. A previous study reported that quercetin 3-rut-7-rha and rutin had hypolipidemic
Jon Bråte et al.
Current biology : CB, 28(20), 3288-3295 (2018-10-16)
The emergence of multicellular animals was associated with an increase in phenotypic complexity and with the acquisition of spatial cell differentiation and embryonic development. Paradoxically, this phenotypic transition was not paralleled by major changes in the underlying developmental toolkit and
Unicellular origin of the animal microRNA machinery
Br?te J, et al.
Current Biology, 28(20), 3288- 3295 (2018)
Abdulla Abdulla Sabana et al.
Planta, 251(4), 79-79 (2020-03-14)
Genome-wide analysis of small RNAs identifies somatic embryogenesis- specific miRNAs and their targets and provides novel insights into the mechanisms governing somatic embryogenesis in coconut, a highly in vitro recalcitrant species. Coconut, a major plantation crop of the tropics is

Articles

Simple DNA/RNA purification methods aid genome analysis from various sources, enhancing research efficiency.

Simple DNA/RNA purification methods aid genome analysis from various sources, enhancing research efficiency.

Simple DNA/RNA purification methods aid genome analysis from various sources, enhancing research efficiency.

Simple DNA/RNA purification methods aid genome analysis from various sources, enhancing research efficiency.

Related Content

Sigma-Aldrich® Advanced Genomics is the leading provider of gene editing and silencing technologies including CRISPR, Cas9, synthetic guide RNA (sgRNA), and Zinc Finger Nuclease (ZFN).

Sigma-Aldrich® Advanced Genomics is the leading provider of gene editing and silencing technologies including CRISPR, Cas9, synthetic guide RNA (sgRNA), and Zinc Finger Nuclease (ZFN).

Sigma-Aldrich® Advanced Genomics is the leading provider of gene editing and silencing technologies including CRISPR, Cas9, synthetic guide RNA (sgRNA), and Zinc Finger Nuclease (ZFN).

Sigma-Aldrich® Advanced Genomics is the leading provider of gene editing and silencing technologies including CRISPR, Cas9, synthetic guide RNA (sgRNA), and Zinc Finger Nuclease (ZFN).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service