Skip to Content
Merck
All Photos(1)

Key Documents

SNC10

Sigma-Aldrich

mirPremier® microRNA Isolation Kit

1 sufficient for 10 preparations

Synonym(s):

microRNA Isolation Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105501
NACRES:
NA.52

usage

1 sufficient for 10 preparations

technique(s)

DNA extraction: suitable

Looking for similar products? Visit Product Comparison Guide

General description

Sigma′s mirPremier microRNA Isolation Kit provides a rapid and efficient method for purifying and enriching miRNAs and other small RNAs from diverse biological sources, including mammalian cell cultures, animal tissues, plant tissues, and microbial cultures, without using hazardous organic extractions. microRNAs (miRNAs) are a class of small RNA molecules, about 21 nucleotides (nt) in length, that regulate gene expression in a variety of manners, including translational repression, mRNA cleavage and deadenylation. In addition, the kit also can be used for isolating total RNA if messenger RNA or other large RNAs are of interest.
.

Application

mirPremier® microRNA Isolation Kit has been used to:
  • prepare small RNA
  • enrich small RNAfractions from the human astrocytes for small RNA sequencing and bioinformatics analysis
  • isolate small RNA from Vigna mungo tissues
  • extract miRNAs to detect the expression of miRNA-16

Features and Benefits

  • Developed to enhance the efficiency of isolating microRNA and other small RNA molecules directly from a wide range of biological samples.
  • Facilitates rapid and effective extraction and concentration of miRNA in 30 minutes for downstream applications.
  • Capable of extracting miRNA with high purity with no detectable large RNA.
  • Hazardous organic extractions are not involved.

Legal Information

mirPremier is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Extraction of Small RNA and qPCR Validation of miRNAs in Vigna mungo
Paul S, et al.
Bio-protocol, 5(5), e1417-e1417 (2015)
Xiaoqiong Tang et al.
Cancer management and research, 12, 2119-2125 (2020-04-14)
LncRNA mortal obligate RNA transcript (MORT) is downregulated in different types of cancer, indicating its involvement in cancer biology. In this study, MORT and miRNA-16 were both downregulated in plasma of mantle cell lymphoma (MCL) patients than that in the
Ying Zhang et al.
Genome biology, 18(1), 105-105 (2017-06-16)
Post-transcriptional regulation of gene expression can be achieved through the control of mRNA stability, cytoplasmic compartmentalization, 3' UTR length and translational efficacy. Spermiogenesis, a process through which haploid male germ cells differentiate into spermatozoa, represents an ideal model for studying
MicroRNAs control mRNA fate by compartmentalization based on 3? UTR length in male germ cells
Zhang Y, et al.
Genome Biology, 18(1), 105-105 (2017)
Identification of microRNA-mRNA regulatory network associated with oxidative DNA damage in human astrocytes
Nwokwu CD, et al.
ASN Neuro, 14, 17590914221101704-17590914221101704 (2022)

Related Content

Sigma-Aldrich® Advanced Genomics is the leading provider of gene editing and silencing technologies including CRISPR, Cas9, synthetic guide RNA (sgRNA), and Zinc Finger Nuclease (ZFN).

Sigma-Aldrich® Advanced Genomics is the leading provider of gene editing and silencing technologies including CRISPR, Cas9, synthetic guide RNA (sgRNA), and Zinc Finger Nuclease (ZFN).

Sigma-Aldrich® Advanced Genomics is the leading provider of gene editing and silencing technologies including CRISPR, Cas9, synthetic guide RNA (sgRNA), and Zinc Finger Nuclease (ZFN).

Sigma-Aldrich® Advanced Genomics is the leading provider of gene editing and silencing technologies including CRISPR, Cas9, synthetic guide RNA (sgRNA), and Zinc Finger Nuclease (ZFN).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service