Skip to Content
Merck

Skip To

SML3183

Grapiprant

≥98% (HPLC), EP4 receptor antagonist , powder

Synonym(s):

2-Ethyl-4,6-dimethyl-1-[4-[2-[[[[(4-methylphenyl)sulfonyl]amino]carbonyl]amino]ethyl]phenyl]-1H-imidazo[4,5-c]pyridine, CJ-023,423, CJ-023423, N-[({2-[4-(2-ethyl-4,6-dimethyl-1H-imidazo [4,5-c]pyridin-1-yl)phenyl]ethyl}amino)carbonyl]-4-methylbenzenesulfonamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C26H29N5O3S
CAS Number:
Molecular Weight:
491.61
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Grapiprant, ≥98% (HPLC)

Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(NS(C1=CC=C(C)C=C1)(=O)=O)NCCC2=CC=C(N3C(CC)=NC4=C3C=C(C)N=C4C)C=C2

InChI

1S/C26H29N5O3S/c1-5-24-29-25-19(4)28-18(3)16-23(25)31(24)21-10-8-20(9-11-21)14-15-27-26(32)30-35(33,34)22-12-6-17(2)7-13-22/h6-13,16H,5,14-15H2,1-4H3,(H2,27,30,32)

InChI key

HZVLFTCYCLXTGV-UHFFFAOYSA-N

Biochem/physiol Actions

Grapiprant (CJ-023423) is an orally available, a potent and selective EP4 receptor antagonist that exhibits significant inhibitory effects on paw swelling, inflammatory biomarkers, synovial inflammation and bone destruction in AIA rats. Grapiprant is highly selective for the human EP4 receptor over other human prostanoid receptor subtypes. It also inhibits PGE2-evoked elevation in intracellular cAMP at the human and rat EP4 receptors with pA2 of 8.3 and 8.2 nM, respectively. In vivo, oral administration of grapiprant significantly reduces thermal hyperalgesia induced by intraplantar injection of PGE2 (ED50 = 12.8 mg/kg). It is also effective in models of acute and chronic inflammatory pain. Grapiprant significantly reduces mechanical hyperalgesia in the carrageenan model. Furthermore, it significantly reverses complete Freund′s adjuvant-induced chronic inflammatory pain response. It produces antihyperalgesic effects in animal models of inflammatory pain. Grapiprant is approved and widely used for treatment of osteoarthritis related pain in dogs. Grapiprant is classify as a non-cyclooxygenase inhibiting non-steroidal anti-inflammatory drug (NSAID).
orally available, a potent and selective EP4 receptor antagonist

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML0877SML0428SML2705
description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

form

powder

form

powder

form

powder

form

powder

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: 10 mg/mL, clear

solubility

DMSO: 5 mg/mL (clear solution)

solubility

DMSO: 2 mg/mL, clear

color

white to beige

color

white to beige

color

white to beige

color

white to beige


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters