Skip to Content
Merck

SML2705

TP-008

≥98% (HPLC)

Synonym(s):

1-[2-(5-Chloro-2-fluorophenyl)-5-methyl-4-pyridinyl]-1,2-dihydro-2-oxo-3H -imidazo[4,5-c ]pyridine-3-acetamide, 2-(1-(2-(5-Chloro-2-fluorophenyl)-5-methylpyridin-4-yl)-2-oxo-1H-imidazo[4,5-c]pyridin-3(2H)-yl)acetamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H15ClFN5O2
CAS Number:
Molecular Weight:
411.82
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

Fc1c(cc(cc1)Cl)c2ncc(c(c2)[n]3c4c([n]([c]3=O)CC(=O)N)cncc4)C

InChI

1S/C20H15ClFN5O2/c1-11-8-25-15(13-6-12(21)2-3-14(13)22)7-17(11)27-16-4-5-24-9-18(16)26(20(27)29)10-19(23)28/h2-9H,10H2,1H3,(H2,23,28)

InChI key

LVEUPFUJRKZPEN-UHFFFAOYSA-N

Biochem/physiol Actions

SGC Frankfurt Pharma donated probe. TP-008 is a cell penetrant, potent and selective ALK4/5 ((activin-like kinase 4/5) dual inhibitor that potently inhibits SMAD2/3 phosphorylation in C2C12 cells. TP-008 exhibits significant cardiac toxicity in mice. It is compound 18 in BMCL 2016 paper.
cell penetrant, potent and selective ALK4/5 dual inhibitor


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library