Skip to Content
Merck
All Photos(1)

Key Documents

SML3097

Sigma-Aldrich

Darapladib

≥98% (HPLC)

Synonym(s):

1-(N-(2-(Diethylamino)ethyl)-N-(4-(4-trifluoromethylphenyl)benzyl)aminocarbonylmethyl)-2-(4-fluorobenzyl)thio-5,6-trimethylenepyrimidin-4-one, N-[2-(Diethylamino)ethyl]-2-[[(4-fluorophenyl)methyl]thio]-4,5,6,7-tetrahydro-4-oxo-N-[[4′-(trifluoromethyl)[1,1′-biphenyl]-4-yl]methyl]-1H-cyclopentapyrimidine-1-acetamide, SB 435445, SB 480848, SB-435445, SB-480848, SB435445, SB480848

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H38F4N4O2S
CAS Number:
Molecular Weight:
666.77
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C1N=C(SCC2=CC=C(F)C=C2)N(CC(N(CCN(CC)CC)CC3=CC=C(C4=CC=C(C(F)(F)F)C=C4)C=C3)=O)C5=C1CCC5

InChI

1S/C36H38F4N4O2S/c1-3-42(4-2)20-21-43(22-25-8-12-27(13-9-25)28-14-16-29(17-15-28)36(38,39)40)33(45)23-44-32-7-5-6-31(32)34(46)41-35(44)47-24-26-10-18-30(37)19-11-26/h8-19H,3-7,20-24H2,1-2H3

InChI key

WDPFJWLDPVQCAJ-UHFFFAOYSA-N

Biochem/physiol Actions

Darapladib (SB-480848) is an orally active and highly potent lipoprotein associated phospholipase A2 inhibitor (rhLp-PLA2 IC50 = 0.25 nM) with little interaction with cytochrome P450 enzymes (3A4 & 2D6 IC50 >26 μM). Darapladib effectively inhibits plasma Lp-PLA2 activtiy ex vivo (by 60% at 10 nM in human plasma & by 79% at 100 nM in Watanabe hereditable hyperlipidaemic (WHHL) rabbit plasma) and in vivo (by 95% & 91%, respectively, 3 & 5 hrs post 10 mg/kg WHHL rabbits via p.o.).
Orally active and highly potent lipoprotein associated phospholipase A2 inhibitor (Lp-PLA2 ) with little interaction with cytochrome P450 enzymes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service