Skip to Content
Merck
All Photos(1)

Key Documents

SML2882

Sigma-Aldrich

NRT-870-59

≥98% (HPLC)

Synonym(s):

7-Imino-?5-?methyl-?2-?phenyl-thieno[3,?2-?c]?pyridine-?4,?6(5H,?7H)?-?dione, 7-imino-5-methyl-2-phenylthieno[3,2-c]pyridine-4,6(5H,7H)-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H10N2O2S
CAS Number:
Molecular Weight:
270.31
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

N=C(C(N1C)=O)C2=C(C=C(C3=CC=CC=C3)S2)C1=O

InChI

1S/C14H10N2O2S/c1-16-13(17)9-7-10(8-5-3-2-4-6-8)19-12(9)11(15)14(16)18/h2-7,15H,1H3

InChI key

IGPZTAJCUQIMOK-UHFFFAOYSA-N

Biochem/physiol Actions

NRT-870-59 is a potent and selective reversible inhibitor of PTP4A3 tyrosine phosphatase that exhibits potent cytotoxicity against human ovarian and breast cancer cells grown as spheroids. NRT-870-59 does not inhibit oncogenic dual specificity phosphatase CDC25B.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nikhil R Tasker et al.
Organic & biomolecular chemistry, 17(9), 2448-2466 (2019-02-13)
A continuous flow photooxygenation of 7-aminothieno[3,2-c]pyridin-4(5H)-ones to produce 7-iminothieno[3,2-c]pyridine-4,6(5H,7H)-diones has been developed, utilizing ambient air as the sole reactant. N-H Imines are formed as the major products, and excellent functional group tolerance and conversion on gram-scale without the need for
John S Lazo et al.
The Journal of pharmacology and experimental therapeutics, 371(3), 652-662 (2019-10-12)
Oncogenic protein tyrosine phosphatases (PTPs) are overexpressed in numerous human cancers but they have been challenging pharmacological targets. The emblematic oncogenic PTP4A tyrosine phosphatase family regulates many fundamental malignant processes. 7-Imino-2-phenylthieno[3,2-c]pyridine-4,6(5H,7H)-dione (JMS-053) is a novel, potent, and selective PTP4A inhibitor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service