If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf
Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
1 kit | Check Cart for Availability | $1,350.00 |
About This Item
usage
sufficient for 100 assays (96 well plates)
Quality Segment
shipped in
wet ice
storage temp.
2-8°C
Application
Biochem/physiol Actions
Analysis Note
1 of 1
This Item | |||
|---|---|---|---|
| Quality Level 200 | Quality Level 200 | Quality Level 200 | Quality Level 200 |
| usage sufficient for 100 assays (96 well plates) | usage sufficient for 100 assays | usage sufficient for 100 assays (96 well plates) | usage sufficient for 700 assays |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. −20°C | storage temp. 2-8°C |
| shipped in wet ice | shipped in - | shipped in dry ice | shipped in wet ice |
Kit Components Only
- Assay Buffer 25 mL
- Assay Medium 20 mL
- Enhancer Solution, 40 mM 500 μL
- Luminol Solution 500 μL
- Xanthine Solution 500 μL
- Xanthine Oxidase .5 U
- Superoxide Dismutase from bovine liver, 4 units/μL 400 U
Storage Class
10 - Combustible liquids
wgk
WGK 1
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
How can I determine the shelf life / expiration / retest date of this product?
1 answer-
Helpful?
-
-
How is shipping temperature determined? And how is it related to the product storage temperature?
1 answer-
Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf
Helpful?
-


