Skip to Content
Merck
All Photos(1)

Key Documents

SML2386

Sigma-Aldrich

Cycloguanil hydrochloride

≥95% (HPLC)

Synonym(s):

1-(4-chlorophenyl)-1,6-dihydro-6,6-dimethyl-1,3,5-triazine-2,4-diamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H14ClN5 · HCl
CAS Number:
Molecular Weight:
288.18
MDL number:
UNSPSC Code:
12352200

Assay

≥95% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

Clc1ccc(cc1)N2C(NC(=N)NC2=N)(C)C.Cl

InChI

1S/C11H14ClN5.ClH/c1-11(2)16-9(13)15-10(14)17(11)8-5-3-7(12)4-6-8;/h3-6H,1-2H3,(H4,13,14,15,16);1H

InChI key

MOUAPRKJJUXEIE-UHFFFAOYSA-N

Application

Cycloguanil hydrochloride is suitable for use as a positive control in screening Plasmodium falciparum strains in microdilution radioisotope antiplasmodial bioassay. It may also be used as internal standard in high performance liquid chromatography for the quantification of urine and plasma samples.

Biochem/physiol Actions

Cycloguanil is it a potential antimalarial drug. It is hydrophilic in nature. Cycloguanil acts as a substrate for the enzyme organic cation transporter 1 (OCT1).
Cycloguanil, a metabolite of proguanil, is a potent dihydrofolate reductase inhibitor.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

OCT 1 Deficiency Affects Hepatocellular Concentrations and Pharmacokinetics of Cycloguanil, the Active Metabolite of the Antimalarial Drug Proguanil
Matthaei J, et al.
Clinical Pharmacology and Therapeutics, 105(1), 190-200 (2019)
GC/GC-MS analysis, isolation and identification of bioactive essential oil components from the Bhutanese medicinal plant, Pleurospermum amabile
Wangchuk P, et al.
Natural Product Communications, 8(9), 1934578X1300800930-1934578X1300800930 (2013)
Sensitive high performance liquid chromatographic method for the determination of proguanil and its metabolites, cycloguanil and 4-chlorophenylbiguanide in biological fluids
Ebeshi BU, et al.
African Journal of Biotechnology, 4(8), 856-861 (2005)
Phenylpropanoids and furanocoumarins as antibacterial and antimalarial constituents of the Bhutanese medicinal plant Pleurospermum amabile
Wangchuk P, et al.
Natural Product Communications, 9(7), 1934578X1400900719-1934578X1400900719 (2014)
Cholwich Nattee et al.
Journal of molecular graphics & modelling, 71, 13-27 (2016-11-12)
Malaria is still one of the most serious diseases in tropical regions. This is due in part to the high resistance against available drugs for the inhibition of parasites, Plasmodium, the cause of the disease. New potent compounds with high

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service