Skip to Content
Merck
All Photos(1)

Key Documents

SML2311

Sigma-Aldrich

BAY 60-7550

≥95% (HPLC)

Synonym(s):

2-(3,4-Dimethoxybenzyl)-7-{(1R)-1-[(1R)-1-hydroxyethyl]-4-phenylbutyl}-5-methyllimidazo[5,1-f][1,2,4]triazin-4(3H)-one, BAY-60−7550, BAY60-7550

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H32N4O4
CAS Number:
Molecular Weight:
476.57
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥95% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

[n]21[nH]c(n[c](c2c(nc1[C@H]([C@H](O)C)CCCc4ccccc4)C)=O)Cc3cc(c(cc3)OC)OC

InChI

1S/C27H32N4O4/c1-17-25-27(33)29-24(16-20-13-14-22(34-3)23(15-20)35-4)30-31(25)26(28-17)21(18(2)32)12-8-11-19-9-6-5-7-10-19/h5-7,9-10,13-15,18,21,32H,8,11-12,16H2,1-4H3,(H,29,30,33)/t18-,21+/m1/s1

InChI key

MYTWFJKBZGMYCS-NQIIRXRSSA-N

Biochem/physiol Actions

BAY 60-7550 is an orally active, potent and selective cGMP-dependent phosphodiesterase PDE2 (PDE2A) inhibitor (human/bovine PDE2 IC50 = 4.7/2.0 nM; bovine PDE1 IC50 = 108 nM, human PDE5/5A/10A/4B IC50 = 240/580/704/940/1830 nM, human PDE3B/7B/8A/9A/11A IC50 >4 μM) with little activity (IC50 >10 μM) toward acetylcholinesterase, mAO-A/B, adenosine deaminase, and many receptor subtypes tested. Bay 60-7550 effectively upregulates cGMP and cAMP level in cultured rat and murine neurons (1 nM-1 μM) exposed to guanylyl cyclase (GC) or adenylyl cyclase (AC) stimulator (1 μM Bay 41-8543 or 2 μM forskolin), respectively, as well as exhibits learning and memory-improving efficacy in rats (0.6-3 mg/kg p.o.) and mice (0.3-1 mg/kg p.o.) in vivo.
Orally active, potent and selective cGMP-dependent phosphodiesterase PDE2 (PDE2A) inhibitor with learning and memory-improving efficacy in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hege E Larsen et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 36(33), 8562-8573 (2016-08-19)
Hypertension is associated with impaired nitric oxide (NO)-cyclic nucleotide (CN)-coupled intracellular calcium (Ca(2+)) homeostasis that enhances cardiac sympathetic neurotransmission. Because neuronal membrane Ca(2+) currents are reduced by NO-activated S-nitrosylation, we tested whether CNs affect membrane channel conductance directly in neurons
Jun-Nan Wang et al.
European spine journal : official publication of the European Spine Society, the European Spinal Deformity Society, and the European Section of the Cervical Spine Research Society, 26(7), 1961-1968 (2017-03-12)
Phosphodiesterase inhibitors possess anti-inflammatory properties. In addition, some studies report that phosphodiesterase 2A (PDE2A) are highly expressed in the dorsal horn of the spinal cord. The present study aimed to investigate whether intrathecal administration of Bay 60-7550, a specific PDE2A
Wen Chen et al.
Circulation research, 119(2), 237-248 (2016-05-05)
In patients after acute myocardial infarction (AMI), the initial extent of necrosis and inflammation determine clinical outcome. One early event in AMI is the increased cardiac expression of atrial natriuretic peptide (NP) and B-type NP, with their plasma levels correlating
Juen Zhang et al.
Cell, 166(3), 716-728 (2016-07-19)
Fear behaviors are regulated by adaptive mechanisms that dampen their expression in the absence of danger. By studying circuits and the molecular mechanisms underlying this adaptive response, we show that cholinergic neurons of the medial habenula reduce fear memory expression
Ligia Mendes Soares et al.
The European journal of neuroscience, 45(4), 510-520 (2016-11-05)
Cognitive and affective impairments are the most characterized consequences following cerebral ischemia. BAY 60-7550, a selective phosphodiesterase type 2 inhibitor (PDE2-I), presents memory-enhancing and anxiolytic-like properties. The behavioral effects of BAY 60-7550 have been associated with its ability to prevent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service