Skip to Content
Merck

CS0720

Citrate Synthase Assay Kit

1 kit sufficient for 100 reactions (using a 1 ml cuvette), 1 kit sufficient for 480 reactions (using 96 multiwell plates)

Synonym(s):

Citrate Synthase Activity Test Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.28
UNSPSC Code:
12161503
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

usage

 kit sufficient for 100 reactions (using a 1 ml cuvette),  kit sufficient for 480 reactions (using 96 multiwell plates)

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... CS(1431)

General description

The Citrate Synthase Assay Kit contains all the required reagents (including a positive control enzyme) for a fast and simple measurement of citrate synthase activity in a whole cell extract or in isolated mitochondria. In addition, the kit enables testing of the intactness of the mitochondrial inner membrane.

Biochem/physiol Actions

Citrate synthase is the initial enzyme of the tricarboxylic acid (TCA) cycle. The enzyme catalyzes the reaction of 2 carbon acetyl CoA with 4 carbon oxaloacetate to form the 6 carbon citrate. This enzyme is an exclusive marker of the mitochondrial matrix.

Analysis Note

The activity of the enzyme is measured by following the color of TNB, which is generated from DTNB present in the reaction of citrate synthesis, and caused by the deacetylation of Acetyl-CoA. The overall reaction product, TNB, absorbs at 412 nm.


Kit Components Only

Product No.
Description

  • Assay buffer for citrate synthase 5x 25 mL

  • Bicine buffer 10 mL

  • CelLytic M Cell Lysis Reagent 10 mL

  • DTNB [5,5'-Dithiobis-(2-nitrobenzoic acid)] 4 mg

  • Acetyl coenzyme A 25 mg

  • Oxaloacetic acid 1 g

  • Citrate synthase (positive control) .1 mL

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library