Skip to Content
Merck

Skip To

SML1887

APX2009

≥98% (HPLC)

Synonym(s):

(2E)-2-[(3-methoxy-1,4-dioxo-1,4-dihydronaphthalen-2-yl)methylidene]-N,N-diethylpentanamide, 2-[(1,4-Dihydro-3-methoxy-1,4-dioxo-2-naphthalenyl)methylene]-N,N-diethyl-pentanamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C21H25NO4
CAS Number:
Molecular Weight:
355.43
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C1C(OC)=C(/C=C(CCC)/C(N(CC)CC)=O)C(C2=C1C=CC=C2)=O

InChI

1S/C21H25NO4/c1-5-10-14(21(25)22(6-2)7-3)13-17-18(23)15-11-8-9-12-16(15)19(24)20(17)26-4/h8-9,11-13H,5-7,10H2,1-4H3/b14-13+

InChI key

WEMXCNYTFZUFSB-BUHFOSPRSA-N

Biochem/physiol Actions

APX2009 is a cell penetrant potent and selective second-generation inhibitor of APE1 redox-signaling that stimulates APE1 DNA repair activity. APX2009 is neuroprotective against cisplatin and oxaliplatin induced toxicity. APX2009 exhibits potent antitumor activity in neuroblastoma cell lines and 3D spheroid pancreatic tumor model.
Cell penetrant potent and selective second-generation inhibitor of APE1 redox-signaling that stimulates APE1 DNA repair activity

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML1055SML2686SML0233
description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

form

powder

form

powder

form

powder

form

powder

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

Quality Level

100

Quality Level

100

Quality Level

-

Quality Level

100

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

2-8°C

solubility

DMSO: 10 mg/mL, clear

solubility

DMSO: 20 mg/mL, clear

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: ≥15 mg/mL

color

white to beige

color

white to beige

color

white to beige

color

faintly yellow to yellow


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters