Skip to Content
Merck
All Photos(1)

Key Documents

SML0233

Sigma-Aldrich

Ro 61-8048

≥98% (HPLC)

Synonym(s):

3,4-Dimethoxy-N-[4-(3-nitrophenyl)-2-thiazolyl]-benzenesulfonamide, 3,4-Dimethoxy-N-[4-(3-nitrophenyl)thiazol-2-yl]benzenesulfonamide, Ro-61-8048

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H15N3O6S2
CAS Number:
Molecular Weight:
421.45
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

faintly yellow to yellow

solubility

DMSO: ≥15 mg/mL

storage temp.

2-8°C

SMILES string

COc1ccc(cc1OC)S(=O)(=O)Nc2nc(cs2)-c3cccc(c3)[N+]([O-])=O

InChI

1S/C17H15N3O6S2/c1-25-15-7-6-13(9-16(15)26-2)28(23,24)19-17-18-14(10-27-17)11-4-3-5-12(8-11)20(21)22/h3-10H,1-2H3,(H,18,19)

InChI key

NDPBMCKQJOZAQX-UHFFFAOYSA-N

Application

Ro 61-8048 has been used as a kynurenine 3-monooxygenase (KMO) inhibitor to study its effects on nicotinamide adenine dinucleotide (NAD+) concentration in hepatocytes of mice.

Biochem/physiol Actions

Inhibition of kynurenine 3-hydroxylase by Ro 61-8048 reduces the episodes of dystonia and dyskinesias induced by decreased levels of kynurenic acid.
Ro 61-8048 is an inhibitor of kynurenine 3-monooxygenase (KMO) that increases kynurenic acid levels and reduces extracellular glutamate in the brain.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jean Rodgers et al.
Brain : a journal of neurology, 132(Pt 5), 1259-1267 (2009-04-03)
Human African trypanosomiasis, or sleeping sickness, is caused by the protozoan parasites Trypanosoma brucei rhodesiense or Trypanosoma brucei gambiense, and is a major cause of systemic and neurological disability throughout sub-Saharan Africa. Following early-stage disease, the trypanosomes cross the blood-brain
Angelika Richter et al.
European journal of pharmacology, 478(1), 47-52 (2003-10-14)
The effects of the novel kynurenine 3-hydroxylase inhibitor 3,4-dimethoxy-N-[4-(3-nitrophenyl)thiazol-2-yl]benzenesulfonamide (Ro 61-8048) on severity of dystonia were examined in dt(sz) mutant hamsters, an animal model of paroxysmal dystonia, in which stress precipitates dystonic episodes. Ro 61-8048 (50, 100 and 150 mg/kg
Arghya Ray et al.
Leukemia, 34(2), 567-577 (2019-08-30)
Our prior studies showed that dysfunctional plasmacytoid dendritic cells (pDCs) contribute to multiple myeloma (MM) pathogenesis. Specifically, pDC interactions with tumor and T/NK effector cells in the bone marrow (BM) milieu induce immune suppression and MM cell proliferation. Delineation of
Melanie Hamann et al.
European journal of pharmacology, 586(1-3), 156-159 (2008-03-21)
Striatal dysfunctions seem to play a key role in the pathophysiology of dystonia in the dt(sz) mutant hamster, a model of paroxysmal non-kinesigenic dyskinesia, in which stress precipitates dystonic episodes. Previous examinations have shown changes in kynurenic acid levels and
Paul B Larkin et al.
Biochimica et biophysica acta, 1860(11 Pt A), 2345-2354 (2016-07-10)
In mammals, the majority of the essential amino acid tryptophan is degraded via the kynurenine pathway (KP). Several KP metabolites play distinct physiological roles, often linked to immune system functions, and may also be causally involved in human diseases including

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service