Skip to Content
Merck
All Photos(1)

Key Documents

SML0674

Sigma-Aldrich

Ramosetron hydrochloride

≥98% (HPLC)

Synonym(s):

(R)-5-[(1-Methyl-3-indolyl)carbonyl]-4,5,6,7-tetrahydro-1H-benzimidazole hydrochloride, YM-060, YM060

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H17N3O · HCl
CAS Number:
Molecular Weight:
315.80
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -36 to -46°, c = 1 in methanol

storage condition

desiccated

color

white to beige

solubility

H2O: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

Cl.[n]1(c2c(c(c1)C(=O)[C@@H]3CCc4nc[nH]c4C3)cccc2)C

InChI

1S/C17H17N3O.ClH/c1-20-9-13(12-4-2-3-5-16(12)20)17(21)11-6-7-14-15(8-11)19-10-18-14;/h2-5,9-11H,6-8H2,1H3,(H,18,19);1H/t11-;/m1./s1

InChI key

XIXYTCLDXQRHJO-RFVHGSKJSA-N

Related Categories

Biochem/physiol Actions

Ramosetron hydrochloride is a potent serotonin 5-HT3 receptor antagonist with higher afinity for the 5-HT3 receptor than other currently available antagonists.
Ramosetron hydrochloride is a potent serotonin 5-HT3 receptor antagonist with higher afinity for the 5-HT3 receptor than other currently available antagonists. Ramosetron is used clinically both as an antiemetic to treat nausea and vomiting following chemotherapy or after surgery, and in treatment of irritable bowel syndrome.
Ramosetron is a tetra hydrobenzimidazole derivative, containing an indole ring. It inhibits colon contraction mediated by serotonin, competitively.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T S Hahm et al.
Anaesthesia, 65(5), 500-504 (2010-03-27)
We compared the prophylactic anti-emetic efficacy of ramosetron, a newly developed 5-HT(3) antagonist, and ondansetron in patients at high-risk for postoperative nausea and vomiting after total knee replacement. Eighty-four patients with three risk factors for postoperative nausea and vomiting (female
Jung-Hee Ryu et al.
Clinical therapeutics, 33(9), 1162-1172 (2011-08-23)
Patients undergoing general anesthesia for laparoscopic cholecystectomy have a high risk of postoperative nausea and vomiting (PONV) with incidences up to 75%. Ramosetron, a serotonin subtype 3 (5-HT(3)) antagonist, has been shown to be effective as an antiemetic after chemotherapy
Serotonin 5-HT3 receptor antagonist for treatment of severe diabetic diarrhea.
Satoshi Murao et al.
Diabetes care, 33(3), e38-e38 (2010-03-02)
Maulana M Ansari et al.
Indian journal of medical sciences, 64(6), 272-280 (2010-06-01)
Post-operative nausea and vomiting (PONV) is common. 5HT 3 receptor antagonists are commonly used drugs for its prevention. A study was designed to compare the efficacy and safety of ramosetron and ondansetron in patients undergoing laparoscopic cholecystectomy (lap chole). A
Takahiro Mihara et al.
Anesthesia and analgesia, 117(2), 329-339 (2013-06-13)
Ramosetron has been shown to have a very strong effect for preventing postoperative nausea and vomiting (PONV) in previous meta-analyses. However, these previous meta-analyses included a number of studies by Fujii et al. which have now been proven to have

Articles

We offer many products related to serotonin receptors for your research needs.

We offer many products related to serotonin receptors for your research needs.

We offer many products related to serotonin receptors for your research needs.

We offer many products related to serotonin receptors for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service