Skip to Content
Merck
All Photos(1)

Key Documents

255653

Sigma-Aldrich

trans-2-Nonenal

97%

Synonym(s):

(2E)-2-Nonenal, (2E)-Non-2-enal, (2E)-Nonen-1-al, (E)-2-Nonen-1-al, (E)-2-Nonenal, (E)-Non-2-enal, (E)-Nonen-1-al, 2-trans-Nonenal, trans-2-Nonen-1-al

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5CH=CHCHO
CAS Number:
Molecular Weight:
140.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

>1 (vs air)

Assay

97%

form

liquid

refractive index

n20/D 1.453 (lit.)

bp

88-90 °C/12 mmHg (lit.)

density

0.846 g/mL at 25 °C (lit.)

functional group

aldehyde

SMILES string

[H]C(=O)C(\[H])=C(/[H])CCCCCC

InChI

1S/C9H16O/c1-2-3-4-5-6-7-8-9-10/h7-9H,2-6H2,1H3/b8-7+

InChI key

BSAIUMLZVGUGKX-BQYQJAHWSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

183.2 °F - closed cup

Flash Point(C)

84 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Galliard et al.
Biochimica et biophysica acta, 441(2), 181-192 (1976-08-23)
1. A particulate enzyme fraction and an acetone powder preparation from cucumber fruits cleaved 9- and 13-hydroperoxyoctadecadienoic acids to form volatile aldehydes and oxoacid fragments. 2. From the 9-hydroperoxide, the major volatile fragments were cis-3-nonenal and trans-2-nonenal using particulate enzyme
Caterina Fattorusso et al.
Bioorganic & medicinal chemistry, 19(1), 312-320 (2010-12-03)
In our search for new antimalarial agents inspired by natural products, we describe herein the synthesis, the evaluation of in vitro antiplasmodial activity, and the SAR studies for a series of endoperoxide antimalarials based on the plakortin scaffold. These simplified
Jan Hubert et al.
Experimental & applied acarology, 44(4), 315-321 (2008-04-19)
The toxicities of three plant volatiles, (2E)-hexenal, (2E, 6Z)-nonadienal and (2E)-nonenal, intermediate products of the oxylipin biosynthesis pathway, were tested on three mites of importance for medical purposes and as pests. The aldehydes were diluted in hexane separately and incorporated
M Spite et al.
British journal of pharmacology, 158(4), 1062-1073 (2009-05-09)
Inflammation is associated with oxidative stress and local generation of lipid peroxidation-derived aldehydes, such as 4-hydroxy-trans-2-nonenal (HNE). In most tissues, HNE is readily conjugated with glutathione and presently it is unknown whether glutathionyl-HNE (GS-HNE) plays a functional role in inflammation.
Giovanna Cenini et al.
Biochimica et biophysica acta, 1822(2), 130-138 (2011-10-20)
Down syndrome (DS) is the most common genetic cause of intellectual disability in children, and the number of adults with DS reaching old age is increasing. By the age of 40 years, virtually all people with DS have sufficient neuropathology

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service