Skip to Content
Merck
All Photos(1)

Key Documents

SML0551

Sigma-Aldrich

Icaritin

≥98% (HPLC)

Synonym(s):

3,5,7-Trihydroxy-2-(4-methoxyphenyl)-8-(3-methyl-2-buten-1-yl)-4H-1-benzopyran-4-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H20O6
CAS Number:
Molecular Weight:
368.38
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

light yellow to dark yellow

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

COc1ccc(cc1)C2=C(O)C(=O)c3c(O)cc(O)c(C\C=C(\C)C)c3O2

InChI

1S/C21H20O6/c1-11(2)4-9-14-15(22)10-16(23)17-18(24)19(25)20(27-21(14)17)12-5-7-13(26-3)8-6-12/h4-8,10,22-23,25H,9H2,1-3H3

InChI key

TUUXBSASAQJECY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Biochem/physiol Actions

Icaritin enhances osteoblastic differentiation of mesenchymal stem cells (MSCs) while it inhibits adipogenic differentiation of MSCs.
Icaritin enhances osteoblastic differentiation of mesenchymal stem cells (MSCs). It may have estrogen-like activities.
Icaritin is a component of Epimedium flavonoid isolated from Herba Epimedii, which enhances osteoblastic differentiation of mesenchymal stem cells (MSCs) while it inhibits adipogenic differentiation of MSCs by inhibiting PPAR-g pathway. Icaritin has no effect on MSCs proliferation. Also, icaritin potently inhibits chronic myeloid leukemia (CML) and breast cancer cells proliferation most likely by modulation of MAPK/ERK/JNK and JAK2/STAT3 /AKT signaling. As other flavonoids, icaritin may exert estrogen-like activities.

Features and Benefits

This compound is featured on the JAKs, MAPKs and Nuclear Receptors (PPARs) pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jian feng Zhu et al.
PloS one, 6(8), e23720-e23720 (2011-09-03)
To explore the effects of Icaritin on chronic myeloid leukemia (CML) cells and underlying mechanisms. CML cells were incubated with various concentration of Icaritin for 48 hours, the cell proliferation was analyzed by MTT and the apoptosis was assessed with
Jing-Shan Tong et al.
PloS one, 6(3), e16781-e16781 (2011-03-17)
Icaritin, a compound from Epimedium Genus, has selective estrogen receptor (ER) modulating activities, and possess anti-tumor activity. Here, we examined icaritin effect on cell growth of human endometrial cancer Hec1A cells and found that icaritin potently inhibited proliferation of Hec1A
Hongchao Li et al.
Journal of cellular biochemistry (2018-11-06)
Glioblastoma (GBM) is a common and aggressive brain tumor that is associated with significant increase in glycolysis for energy production. Icaritin is a natural compound and exhibits anticancer activity in GBM. However, the effect of icaritin on glycolysis in GBM
Dong Yao et al.
PloS one, 7(8), e41264-e41264 (2012-09-07)
We found that Icaritin, an intestinal metabolite of Epimedium-derived flavonoids (EF) enhanced osteoblastic differentiation of mesenchymal stem cells (MSCs) only under osteogenic induction conditions. We also demonstrated its effect on inhibition of adipogenic differentiation of MSCs. Unlike the findings of
Haibang Hao et al.
European journal of immunology, 49(12), 2235-2244 (2019-08-30)
Icaritin, a hydrolytic product of icariin isolated from traditional Chinese herbal medicine genus Epimedium, has many pharmacological and biological activities. Here, we show that icaritin can effectively decrease tumor burden of murine B16F10 melanoma and MC38 colorectal tumors in a

Articles

Peroxisome proliferator activated receptors (PPARs) are ligand-activated transcription factors related to hormone receptors, influencing gene expression.

Peroxisome proliferator activated receptors (PPARs) are ligand-activated transcription factors related to hormone receptors, influencing gene expression.

Peroxisome proliferator activated receptors (PPARs) are ligand-activated transcription factors related to hormone receptors, influencing gene expression.

Peroxisome proliferator activated receptors (PPARs) are ligand-activated transcription factors related to hormone receptors, influencing gene expression.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service