42575
Dehydrocostus lactone
analytical standard
Synonym(s):
(−)-Dehydrocostus lactone; Dehydrocostus lactone; Epiligulyl oxide, (3aS,6aR,9aR,9bS)-Decahydro-3,6,9-tris(methylene)-azuleno[4,5-b]furan-2(3H)-one, Dehydro-costus lactone (6CI,7CI), Dehydrocostuslactone, Guaia-4(15),10(14),11(13)-trien-12-oic acid, 6alpha-hydroxy-, gamma-lactone, [3aS-(3aα,6aα,9aα,9bβ)]-Decahydro-3,6,9-tris(methylene)-azuleno[4,5-b]furan-2(3H)-one
About This Item
grade
analytical standard
Quality Level
Assay
≥98.0% (GC)
optical activity
[α]/D -14.0±3.0°, c = 1 in chloroform
shelf life
limited shelf life, expiry date on the label
impurities
≤2.0% water
format
neat
storage temp.
−20°C
SMILES string
C=C1CC[C@@]2([H])C(CC[C@@H]3[C@H](OC(C3=C)=O)[C@]21[H])=C
InChI
1S/C15H18O2/c1-8-4-7-12-10(3)15(16)17-14(12)13-9(2)5-6-11(8)13/h11-14H,1-7H2/t11-,12-,13-,14-/m0/s1
InChI key
NETSQGRTUNRXEO-XUXIUFHCSA-N
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Sens. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service