Skip to Content
Merck
All Photos(1)

Key Documents

PHR1097

Supelco

L-Tyrosine

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

(S)-2-Amino-3-(4-hydroxyphenyl)propionic acid, 3-(4-Hydroxyphenyl)-L-alanine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(HO)C6H4CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
181.19
Beilstein:
392441
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. T2900000
traceable to USP 1705006

API family

tyrosine

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

>300 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

N[C@@H](Cc1ccc(O)cc1)C(O)=O

InChI

1S/C9H11NO3/c10-8(9(12)13)5-6-1-3-7(11)4-2-6/h1-4,8,11H,5,10H2,(H,12,13)/t8-/m0/s1

InChI key

OUYCCCASQSFEME-QMMMGPOBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

L-Tyrosine may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by a liquid chromatography technique.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Other Notes

Amino acid precursor of dopamine and other catecholamines
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA7163 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High-performance liquid chromatographic analysis of biogenic amines in pharmaceutical products containing Citrus aurantium
Viana C, et al.
Food Additives & Contaminants. Part A, Chemistry, Analysis, Control, Exposure & Risk Assessment, 30(4), 634-642 (2013)
T Eklundh et al.
Journal of psychiatric research, 30(3), 201-208 (1996-05-01)
In order to elucidate methodological aspects of CSF investigations, 14 healthy male subjects were lumbar-punctured at the L4-5 level following a standardised procedure. CSF concentrations of precursors, transmitters and transmitter metabolites were used as dependent variables, while age, height, body
Alena Volgusheva et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(18), 7223-7228 (2013-04-17)
Photobiological H2 production is an attractive option for renewable solar fuels. Sulfur-deprived cells of Chlamydomonas reinhardtii have been shown to produce hydrogen with the highest efficiency among photobiological systems. We have investigated the photosynthetic reactions during sulfur deprivation and H2
Vincent Dunet et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 53(2), 207-214 (2012-02-04)
For the past decade, PET with (18)F-fluoro-ethyl-tyrosine ((18)F-FET) has been used in the evaluation of patients with primary brain tumors (PBTs), but so far series have reported only a limited number of patients. The purpose of this systematic review and
Verena Rahm et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 55(4), 546-550 (2014-02-26)
Low-grade gliomas (LGGs) may harbor malignant foci, which are characterized by increased tumor cellularity and angiogenesis. We used diffusion-weighted MR imaging (apparent diffusion coefficient [ADC]) and PET with the amino acid O-(2-(18)F-fluorethyl)-L-tyrosine ((18)F-FET) to search for focal changes of diffusion

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service