Skip to Content
Merck
All Photos(1)

Documents

SMB00092

Sigma-Aldrich

Osajin

≥95% (LC/MS-ELSD)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H24O5
CAS Number:
Molecular Weight:
404.46
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Assay

≥95% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

C\C(C)=C\Cc1c(O)c2C(=O)C(=COc2c3C=CC(C)(C)Oc13)c4ccc(O)cc4

InChI

1S/C25H24O5/c1-14(2)5-10-17-21(27)20-22(28)19(15-6-8-16(26)9-7-15)13-29-24(20)18-11-12-25(3,4)30-23(17)18/h5-9,11-13,26-27H,10H2,1-4H3

InChI key

DCTLJGWMHPGCOS-UHFFFAOYSA-N

General description

Natural product derived from plant source.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Bartosíková et al.
Die Pharmazie, 61(6), 552-555 (2006-07-11)
The aim of this study was to analyze the antioxidative effect of osajin during prophylactic administration. The pathological model for in vivo experiment was the unilateral ischemia-reperfusion of kidney of the laboratory rat. The animals were randomly divided into five
James V Gruber et al.
Mediators of inflammation, 2010, 230450-230450 (2010-08-14)
A series of well-known, purified antioxidants including: Resveratrol, Epigallocatechin Gallate (EGCG), Genistein, Rosavin, Puerarin, Chlorogenic Acid, Propolis and two newer unexplored isoflavonoids isolated from Maclura pomifera (Osage Orange) including Pomiferin and Osajin, were applied to Normal Human Dermal Fibroblasts (NHDF)
Tsung-Teng Huang et al.
PloS one, 6(4), e18308-e18308 (2011-05-03)
Osajin is a prenylated isoflavone showing antitumor activity in different tumor cell lines. The underlying mechanism of osajin-induced cancer cell death is not clearly understood. In the present study, the mechanisms of osajin-induced cell death of human nasopharyngeal carcinoma (NPC)
William L Whaley et al.
Langmuir : the ACS journal of surfaces and colloids, 22(17), 7175-7184 (2006-08-09)
Genistein (5,7,4'-trihydroxyisoflavone) modulates the function of several transmembrane ion-channel proteins by mechanisms that are unrelated to phosphorylation events. Daidzein (7,4'-dihydroxy-isoflavone) typically exhibits modest effects, whereas genistin (7-O-glucosyl-genistein) usually exhibits no effect on ion-channel activities. Genistein appears to modulate gramicidin A
Il Hong Son et al.
Bioorganic & medicinal chemistry letters, 17(17), 4753-4755 (2007-07-31)
The major constituents from the fruits of Maclura pomifera are the prenylated isoflavones, osajin (1) and pomiferin (2). Their structures were elucidated using NMR spectroscopic techniques and mass spectrometric analysis. Compound 2 showed potential inhibitory activity in histone deacetylase (HDAC)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service