Skip to Content
Merck
All Photos(1)

Documents

SMB00088

Sigma-Aldrich

Oregonin

≥95% (LC/MS-ELSD)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H30O10
CAS Number:
Molecular Weight:
478.49
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Assay

≥95% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

OC1=CC=C(CCC(C[C@H](CCC2=CC(O)=C(O)C=C2)O[C@H]3[C@H](O)[C@@H](O)[C@H](O)CO3)=O)C=C1O

InChI

1S/C24H30O10/c25-15(5-1-13-3-7-17(26)19(28)9-13)11-16(6-2-14-4-8-18(27)20(29)10-14)34-24-23(32)22(31)21(30)12-33-24/h3-4,7-10,16,21-24,26-32H,1-2,5-6,11-12H2/t16-,21+,22-,23+,24-/m0/s1

InChI key

AQRNEKDRSXYJIN-IRFILORWSA-N

General description

Natural product derived from plant source.

Biochem/physiol Actions

Oregonin is a diarylheptanoid with known antioxidant, anti-cancer, and anti-inflammatory properties.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ching-Hua Kuo et al.
Journal of pharmaceutical and biomedical analysis, 47(1), 195-200 (2008-02-05)
Oregonin, existing primarily in the Alnus plants, displayed anti-inflammatory and antioxidative activities. The capillary zone electrophoresis (CZE) method was developed in this study to quantitatively determine oregonin content in the Alnus plants for the first time. Various parameters, including buffer
Myung Joo Kang et al.
Biological & pharmaceutical bulletin, 33(1), 100-106 (2010-01-05)
To develop an external preparation of oregonin (ORG) for the treatment of atopic dermatitis (AD), conventional creams (CC) and elastic liposomes (EL) containing ORG have been formulated and examined for their in vitro skin permeation properties and in vivo therapeutic
Louis C Martineau et al.
Planta medica, 76(14), 1519-1524 (2010-03-20)
Among modern day metabolic diseases, obesity has reached epidemic proportions worldwide and novel therapeutic support strategies are urgently needed. Adipocytes are interesting targets in this context. Using ethnobotanical and bioassay screening techniques, we have identified two Boreal Forest plants used
Seong-Soo Joo et al.
Archives of pharmacal research, 25(4), 493-499 (2002-09-07)
We investigated the antitumor activity of oregonin, a diarylheptanoid derivative purified from Alnus hirsuta Turcz, Betulaceae. Oregonin is a potential novel immunomodulator, which augments the activation of natural killer (NK) cells, and thereby leads to a powerful antitumor activity. To
Sara L Jackrel et al.
Oecologia, 186(4), 895-906 (2018-02-27)
Differences among individuals within species affect community and ecosystem processes in many systems, and may rival the importance of differences between species. Intraspecific variation consists of both plastic and genetic components that are regulated by different processes and operate on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service